Skip to Content
Merck
All Photos(8)

Key Documents

793612

Sigma-Aldrich

Magnesium sulfate

anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
reagent grade
form:
powder or granules
Pricing and availability is not currently available.

grade

anhydrous
reagent grade

Quality Level

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

Redi-Dri

Assay

≥97%

form

powder or granules

quality

free-flowing

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium sulfate is an inorganic salt that is used as a magnesium precursor to synthesize Mg-based compounds. It is also used as a catalyst for various organic transformations.

Application

Magnesium sulfate can be used as a precursor salt to synthesize magnesium manganese spinel (MgMn2O4) as cathode material for use in rechargeable Mg batteries. It is also used in aqueous two-phase partitioning systems along with polyethylene glycol for the purification of biomolecules.
MgSO4 (anhydrous) can also be used as a drying agent in organic synthesis for the removal of traces of water from organic extracts.

Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders      
  • Safety – Eliminates tools of force used to break up clumps,· 
  • Ecology – Reduces waste as it does not stick to the packaging
  • Economy – Faster preparation and solvation increase productivity and reduce costs     
  • Assured Quality – Excellent, expert-tested quality with no anti-caking agents     
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J Floor et al.
Chembiochem : a European journal of chemical biology, 15(11), 1660-1672 (2014-07-01)
We explored the use of a computational design framework for the stabilization of the haloalkane dehalogenase LinB. Energy calculations, disulfide bond design, molecular dynamics simulations, and rational inspection of mutant structures predicted many stabilizing mutations. Screening of these in small
Claire L O'Brien et al.
Gut, 63(10), 1596-1606 (2014-01-17)
Our aim was to determine whether or not specific microorganisms were transported selectively to lymph nodes in Crohn's disease (CD) by comparing node and mucosal microbial communities in patients and controls. We also sought evidence of dysbiosis and bacterial translocation.
Federica Belluti et al.
ChemMedChem, 9(2), 371-382 (2014-01-10)
A significant improvement in the treatment of trypanosomiases has been achieved with the recent development of nifurtimox-eflornithine combination therapy (NECT). As an alternative to drug combinations and as a means to overcome most of the antitrypanosomatid drug discovery challenges, a
Roberto J Diaz et al.
American journal of physiology. Cell physiology, 306(12), C1191-C1199 (2014-04-25)
We have previously shown that ischemic preconditioning (IPC) protection against necrosis in whole hearts and in both fresh and cultured cardiomyocytes, as well as the improved regulatory volume decrease to hypoosmotic swelling in cardiomyocytes, is abrogated through Cl(-) channel blockade
Audrey Beaussart et al.
ACS nano, 8(10), 10723-10733 (2014-10-07)
A variety of bacterial pathogens use nanoscale protein fibers called type IV pili to mediate cell adhesion, a primary step leading to infection. Currently, how these nanofibers respond to mechanical stimuli and how this response is used to control adhesion

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service