Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium sulfate
Synonym(s):
Magnesium sulfate anhydrous
Empirical Formula (Hill Notation):
MgSO
4
CAS Number:
7487-88-9
Molecular Weight:
120.37
EC Number:
231-298-2
Products
Product Category
salts (8)
Brand
Sigma-Aldrich (7)
Supelco (1)
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
Anhydrous (5)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
M7506
Magnesium sulfate,
anhydrous,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
208094
Magnesium sulfate,
anhydrous, reagent grade, ≥97%
Expand
View Pricing
746452
Magnesium sulfate,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
M2643
Magnesium sulfate,
BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
63136
Magnesium sulfate,
puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Expand
View Pricing
1.06067
Magnesium sulfate,
anhydrous for analysis EMSURE
®
Expand
View Pricing
793612
Magnesium sulfate,
anhydrous, free-flowing, Redi-Dri
™
, reagent grade, ≥97%
Expand
View Pricing
203726
Magnesium sulfate,
≥99.99% trace metals basis
Expand
View Pricing