Merck
  • Enhanced cell volume regulation: a key mechanism in local and remote ischemic preconditioning.
Enhanced cell volume regulation: a key mechanism in local and remote ischemic preconditioning.
American journal of physiology. Cell physiology (2014-04-25)
Roberto J Diaz, Kordan Harvey, Azadeh Boloorchi, Taneya Hossain, Alina Hinek, Peter H Backx, Gregory J Wilson
ABSTRACT
We have previously shown that ischemic preconditioning (IPC) protection against necrosis in whole hearts and in both fresh and cultured cardiomyocytes, as well as the improved regulatory volume decrease to hypoosmotic swelling in cardiomyocytes, is abrogated through Cl(-) channel blockade, pointing to a role for enhanced cell volume regulation in IPC. To further define this cardioprotective mechanism, cultured rabbit ventricular cardiomyocytes were preconditioned either by 10-min simulated ischemia (SI) followed by 10-min simulated reperfusion (SR), by 10-min exposure/10-min washout of remote IPC (rIPC) plasma dialysate (from rabbits subjected to repetitive limb ischemia), or by adenoviral transfection with the constitutively active PKC-ε gene. These interventions were done before cardiomyocytes were subjected to either 60- or 75-min SI/60-min SR to assess cell necrosis (by trypan blue staining), 30-min SI to assess ischemic cell swelling, or 30-min hypoosmotic (200 mosM) stress to assess cell volume regulation. Necrosis after SI/SR and both SI- and hypoosmotic stress-induced swelling was reduced in preconditioned cardiomyocytes compared with control cardiomyocytes (neither preconditioned nor transfected). These effects on necrosis and cell swelling were blocked by either Cl(-) channel blockade or dominant negative knockdown of inwardly rectifying K(+) channels with adenoviruses, suggesting that Cl(-) and K(+) movements across the sarcolemma are critical for cell volume regulation and, thereby, cell survival under hypoxic/ischemic conditions. Our results define enhanced cell volume regulation as a key common mechanism of cardioprotection by preconditioning in cardiomyocytes.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)