Skip to Content
Merck
All Photos(2)

Documents

398098

Sigma-Aldrich

Hydriodic acid

contains no stabilizer, ACS reagent, 55%

Synonym(s):

Hydriotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HI
CAS Number:
Molecular Weight:
127.91
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

55%
55.0-58.0% (ACS specification)

form

liquid

does not contain

stabilizer

impurities

≤0.75% I2

ign. residue

≤0.01%

bp

127 °C (lit.)

density

1.7 g/mL at 25 °C

anion traces

bromide, chloride (as Cl-): ≤0.05%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

storage temp.

2-8°C

SMILES string

I

InChI

1S/HI/h1H

InChI key

XMBWDFGMSWQBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydriodic acid, a strong acid, is hydrogen iodide gas dissolved in water.

Application

Hydriodic acid was used for derivatization of urine samples by iodination to detect aromatic amines using multi-dimensional gas chromatography mass spectrometry (GCxGC-qMS).
It may be used in the following processes:
  • Deiodination process during the synthesis of partially fluorinated pyridinium bromides.
  • As a reducing agent for oxidized graphene nanoribbons (OGN).
  • In combination with red phosphorus to reduce pseudoephedrine in the synthesis of methamphetamine.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graphite oxide and graphene nanoribbons reduction with hydrogen iodide.
Cataldo F, et al.
Fullerenes, Nanotubes, and Carbon Nanostructures, 19(5), 461-468 (2011)
Patnaik P.
A Comprehensive Guide to the Hazardous Properties of Chemical Substances, 123-123 (2007)
Synthesis and biocompatibility evaluation of partially fluorinated pyridinium bromides.
Vyas SM, et al.
New. J. Chem., 30(6), 944-951 (2006)
Relationships between circulating and intracellular thyroid hormones: physiological and clinical implications.
P R Larsen et al.
Endocrine reviews, 2(1), 87-102 (1981-01-01)
Kazumasa Wakamatsu et al.
Pigment cell & melanoma research, 22(4), 474-486 (2009-06-06)
Pheomelanogenesis is a complex pathway that starts with the oxidation of tyrosine (or DOPA, 3,4-dihydroxyphenylalanine) by tyrosinase in the presence of cysteine, which results in the production of 5-S-cysteinyldopa and its isomers. Beyond that step, relatively little has been clarified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service