8.43955
2-Iodopropane
(stabilised) for synthesis
Synonym(s):
2-Iodopropane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H7I
CAS Number:
Molecular Weight:
169.99
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-859-3
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥98.0% (GC)
mp
-90 °C
transition temp
flash point 42 °C
density
1.703 g/cm3 at 25 °C
storage temp.
2-30°C
InChI
1S/C3H7I/c1-3(2)4/h3H,1-2H3
InChI key
FMKOJHQHASLBPH-UHFFFAOYSA-N
Analysis Note
Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.680 - 1.720
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 1.680 - 1.720
Identity (IR): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
107.6 °F - closed cup
Flash Point(C)
42 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service