Skip to Content
Merck
All Photos(3)

Documents

248649

Sigma-Aldrich

Hydriodic acid

contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%

Synonym(s):

Hydriotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HI
CAS Number:
Molecular Weight:
127.91
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

liquid

contains

≤1.5% hypophosphorous acid as stabilizer

concentration

≥47.0%

bp

127 °C (lit.)

density

1.5 g/mL at 25 °C

anion traces

bromide, chloride (as Cl-): ≤0.05%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals: ≤0.001%

storage temp.

2-8°C

SMILES string

I

InChI

1S/HI/h1H

InChI key

XMBWDFGMSWQBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydriodic acid is a solution of hydrogen iodide in water. It is a strong acid. It is commonly used as a reagent in various reactions such as:
  • electrophilic hydriodination of alkenes and alkynes
  • cleavage of epoxides, ethers, and acetals; conversion of alcohols to iodides   
  • reducing agent for many groups including quinones, α-diketones, α-ketols, α-halo ketones, α-diazo ketones, and sulfoxides      
  • reductive cyclization of keto acids

Hydriodic acid (hypophosphorus acid as a stabilizer) can be used for the fabrication of benzylammonium hybrid layered metal-halide perovskites for spintronic and nonlinear optical applications

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siegfried Eigler et al.
Chemical communications (Cambridge, England), 49(67), 7391-7393 (2013-07-19)
The efficiency of reducing agents for the reduction of graphene oxide (GO) could be probed by scanning Raman spectroscopy. A film of graphene flakes derived from GO was probed to be graphene like. We also focus on the surface quality
Viscosities of aqueous hydrochloric acid solutions, and densities and viscosities of aqueous hydroiodic acid solutions.
Nishikata E, et al.
Journal of Chemical and Engineering Data, 26(3), 254-256 (1981)
Efficient reduction of polycyclic quinones, hydroquinones, and phenols with hydriodic acid.
Konieczny M and Harvey RG.
The Journal of Organic Chemistry, 44(26), 4813-4816 (1979)
Relationships between circulating and intracellular thyroid hormones: physiological and clinical implications.
P R Larsen et al.
Endocrine reviews, 2(1), 87-102 (1981-01-01)
Kazumasa Wakamatsu et al.
Pigment cell & melanoma research, 22(4), 474-486 (2009-06-06)
Pheomelanogenesis is a complex pathway that starts with the oxidation of tyrosine (or DOPA, 3,4-dihydroxyphenylalanine) by tyrosinase in the presence of cysteine, which results in the production of 5-S-cysteinyldopa and its isomers. Beyond that step, relatively little has been clarified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service