Skip to Content
Merck
All Photos(1)

Key Documents

38840

Sigma-Aldrich

N,N-Dimethylacetamide

puriss. p.a., ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
164.5-166 °C (lit.)
vapor pressure:
2 mmHg ( 25 °C)
4 mmHg ( 38 °C)
Pricing and availability is not currently available.

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

technique(s)

GC/GC: suitable

impurities

≤0.1% water

evapn. residue

≤0.005%

refractive index

n20/D 1.437 (lit.)
n20/D 1.437

pH

4 (20 °C, 200 g/L)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacetamide (DMAc) is widely used to compose the solvent mixtures for cellulose. Static and dynamic light scattering studies of the cellulose dissolved in varying concentrations of DMAc/LiCl mixtures have been reported.[1]

Application

N,N-Dimethylacetamide (DMAc) may be used to compose the solvent system along with lithium chloride (LiCl) for the dissolution of commercial rayon grade cellulose.[2] DMAc/LiCl solvent mixtures have been used for the preparation of chitin, cellulose and polyimide fibers and films.[3]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Tosh et al.
Carbohydrate research, 327(3), 345-352 (2000-08-17)
Commercial rayon grade cellulose was dissolved in the lithium chloride-N,N-dimethylacetamide (LiCl-DMAc) solvent system and esterified with acetic anhydride using p-toluenesulfonyl chloride (p-TsCl) and pyridine as catalysts. The reaction temperature was varied from 28 to 70 degrees C and the time
Solutions of cellulose in N, N-dimethylacetamide/lithium chloride studied by light scattering methods.
Roder T, et al.
Polymer, 42(16), 6765-6773 (2001)
Solution studies of cellulose in lithium chloride and N,N-dimethylacetamide.
McCormick CL, et al.
Macromolecules, 18(12), 2394-2401 (1985)
Fen Ran et al.
Acta biomaterialia, 7(9), 3370-3381 (2011-06-11)
An amphiphilic triblock co-polymer of poly(vinyl pyrrolidone)-b-poly(methyl methacrylate)-b-poly(vinyl pyrrolidone) (PVP-b-PMMA-b-PVP) was synthesized via reversible addition-fragmentation chain transfer (RAFT) polymerization. The block co-polymer can be directly blended with polyethersulfone (PES) using dimethylacetamide (DMAC) as the solvent to prepare flat sheet and
André H Gröschel et al.
Nature communications, 3, 710-710 (2012-03-20)
Hierarchical self-assembly offers elegant and energy-efficient bottom-up strategies for the structuring of complex materials. For block copolymers, the last decade witnessed great progress in diversifying the structural complexity of solution-based assemblies into multicompartment micelles. However, a general understanding of what

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service