Skip to Content
Merck

Skip To

1.06549

Sodium nitrite

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium nitrite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
NNaO2
CAS Number:
Molecular Weight:
69.00
UNSPSC Code:
12352302
EC Index Number:
231-555-9
NACRES:
NA.21
MDL number:
Assay:
≥99.0% (manganometric)
Grade:
ACS reagent
Form:
solid
Solubility:
820 g/L
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

product line

EMSURE®

assay

≥99.0% (manganometric)

form

solid

potency

85 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

9 (20 °C, 100 g/L in H2O)

mp

280 °C (decomposition)

solubility

820 g/L

density

2.1 g/cm3 at 20 °C

bulk density

1200 kg/m3

anion traces

chloride (Cl-): ≤0.005%, sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.002%, Fe: ≤0.001%, K: ≤0.001%, heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].N(=O)[O-]

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Analysis Note

Assay (manganometric): ≥ 99.0 %
Insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.001 %
Ca (Calcium): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.001 %
Corresponds to ACS, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
6739831443237213
description

for analysis EMSURE® ACS,Reag. Ph Eur

description

analytical standard

description

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%

description

ACS reagent, ≥97.0%

form

solid

form

-

form

powder or crystals

form

powder or crystals

solubility

820 g/L

solubility

-

solubility

-

solubility

-

assay

≥99.0% (manganometric)

assay

≥99.999% trace metals basis, 98.0-102.0% (RT)

assay

≥99%

assay

≥97.0%

Quality Level

300

Quality Level

100

Quality Level

200

Quality Level

200

grade

ACS reagent

grade

analytical standard

grade

ACS reagent, puriss. p.a.

grade

ACS reagent

mp

280 °C (decomposition)

mp

271 °C (lit.)

mp

271 °C (lit.)

mp

271 °C (lit.)


Still not finding the right product?

Explore all of our products under Sodium nitrite


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters