Skip to Content
Merck
All Photos(1)

Key Documents

309079

Sigma-Aldrich

Nitric acid

red, fuming, HNO3 >90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

8 mmHg ( 20 °C)

Quality Level

Assay

5.0-10.0% (% N2O3, titration)

form

liquid

composition

HNO3, >90%

impurities

5-10% dissolved oxides as N2O3

color

red

pH

<1 (20 °C)

bp

120.5 °C (lit.)

density

1.48 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nitric acid is an oxidizing agent used in the oxidation and nitration of organic molecules.

Application

Nitric acid can be used as:
  • A catalyst for the aerobic conversion of benzyl ethers to benzaldehydes.
  • An oxidant for the oxidative bromination of activated aromatic compounds using alkali metal bromide salts to synthesize corresponding bromo-derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 590-590 (1994)
Eduardo Carasek et al.
Talanta, 56(1), 185-191 (2008-10-31)
The need for highly reliable methods for the determination of trace elements has been recognised in analytical chemistry and environmental science. A method for the trace analysis of Pb and Cd in natural waters is described. In a preconcentration step
Heterogeneous reactivity of gaseous nitric acid on Al2O3, CaCO3, and atmospheric dust samples: A Knudsen cell study.
Hanisch F and Crowley JN.
The Journal of Physical Chemistry A, 105(13), 3096-3106 (2001)
Nitric acid purification of single-walled carbon nanotubes.
Hu H, et al.
The Journal of Physical Chemistry B, 107(5), 13838-13842 (2003)
Oxidation of multiwalled carbon nanotubes by nitric acid.
Rosca ID, et al.
Carbon, 43(15), 3124-3131 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service