Skip to Content
Merck
All Photos(3)

Key Documents

431362

Sigma-Aldrich

Hydroxylamine hydrochloride

99.999% trace metals basis

Synonym(s):

Hydroxylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49
Beilstein:
3539763
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22
grade:
ACS reagent (specifications)

grade

ACS reagent (specifications)

Quality Level

Assay

99.999% trace metals basis

form

crystals

impurities

≤0.005% S compounds
≤0.25 meq/g Titr. free acid
<10 ppm total metallic impurities

ign. residue

≤0.05%

pH

2.5-3.5 (20 °C, 50 g/L)

mp

155-157 °C (dec.) (lit.)

density

1.67 g/mL at 25 °C (lit.)

cation traces

Fe: ≤5 ppm
NH4+:, passes test
heavy metals: ≤5 ppm

SMILES string

Cl.NO

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

InChI key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Organosilane amines as potent inhibitors and structural probes of influenza A virus M2 proton channel
  • Lamellarin D analogues as inibitors of topoisomerase I and potential antitumor agents
  • Azapeptide tocolytic agents as inhibitors of prostaglandin F2a receptor for preventing preterm labor
  • Thiazolidinones spiro-fused to indolin-2-ones as potent and selective inhibitors of Mycobacterium tuberculosis protein tyrosine phosphatase B
  • Orally bioavailable quinoline-based antidiabetic dipeptidyl peptidase IV inhibitors targeting Lys554
  • Pyrimidine nucleoside derivatives with nitric oxide donors as antiviral agents
  • Benzyladenosine compounds targeting adenosine A2A receptor and adenosine transporter for neuroprotection
  • Naphthol derivatives as inhibitors of the vanilloid receptor TRPV1 with improved potency in rat cystometry models of urinary incontinence

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Features and Benefits

Meets A.C.S. reagent specifications.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

spleen

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lizethly Cáceres-Jensen et al.
International journal of environmental research and public health, 18(12) (2021-07-03)
The sorption behavior of 2,4-dichlorophenoxyacetic acid (2,4-D) in the abundant agricultural volcanic ash-derived soils (VADS) is not well understood despite being widely used throughout the world, causing effects to the environment and human health. The environmental behavior and risk assessment
Klaus Urbahns et al.
Bioorganic & medicinal chemistry letters, 21(11), 3354-3357 (2011-05-03)
We have identified naphthol derivatives as inhibitors of the vanilloid receptor TRPV1 by high throughput screening. The initial lead showed high clearance in rats and has been optimized by enhancing the acidity of the phenol group. Compound 6b has reduced
Carine B Bourguet et al.
Journal of medicinal chemistry, 54(17), 6085-6097 (2011-07-22)
The prostaglandin-F2α (PGF2α) receptor (FP) was targeted to develop tocolytic agents for inhibiting preterm labor. Azabicycloalkane and azapeptide mimics 2-10 were synthesized based on the (3S,6S,9S)-indolizidin-2-one amino acid analogue PDC113.824 (1), which was shown to modulate FP by a biased
Hironobu Maezaki et al.
Bioorganic & medicinal chemistry, 19(15), 4482-4498 (2011-07-12)
Dipeptidyl peptidase IV (DPP-4) inhibition is a validated therapeutic option for type 2 diabetes, exhibiting multiple antidiabetic effects with little or no risk of hypoglycemia. In our studies involving non-covalent DPP-4 inhibitors, a novel series of quinoline-based inhibitors were designed
Jun Wang et al.
Journal of the American Chemical Society, 133(35), 13844-13847 (2011-08-09)
We describe the use of organosilanes as inhibitors and structural probes of a membrane protein, the M2 proton channel from influenza A virus. Organosilane amine inhibitors were found to be generally as potent as their carbon analogues in targeting WT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service