Skip to Content
Merck
All Photos(2)

Documents

256374

Sigma-Aldrich

2-Ethoxyethanol

spectrophotometric grade, ≥99%

Synonym(s):

Ethyl glycol, Ethylene glycol monoethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098271
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21

grade

spectrophotometric grade

Quality Level

vapor density

3.1 (vs air)

vapor pressure

3.8 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

460 °F

expl. lim.

14 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.080% water

evapn. residue

<0.001%

refractive index

n20/D 1.407 (lit.)

bp

135 °C (lit.)
135 °C

mp

−90 °C (lit.)

solubility

water: miscible

density

0.93 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 215 nm Amax: 1.00
λ: 225 nm Amax: 0.50
λ: 250 nm Amax: 0.20
λ: 305 nm Amax: 0.01

SMILES string

CCOCCO

InChI

1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3

InChI key

ZNQVEEAIQZEUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of polymerizable 1, 8-naphthalimide dyes containing hindered amine fragment.
Bojinov Vl and Konstantinova T.
Dyes and Pigments, 54(3), 239-245 (2003)
Measurements and correlation of high-pressure VLE of binary CO2-alcohol systems (methanol, ethanol, 2-methoxyethanol and 2-ethoxyethanol).
Joung SN, et al.
Fluid Phase Equilibria, 185(1), 219-230 (2001)
Density, Viscosity, Refractive Index, and Speed of Sound in Binary Mixtures of 2-Ethoxyethanol with n-Alkanes (C6 to C12), 2, 2, 4-Trimethylpentane, and Cyclohexane in the Temperature Interval 298.15-313.15 K.
Aminabhavi TM and Gopalakrishna B.
Journal of Chemical and Engineering Data, 40(3), 632-641 (1995)
Sanne A B Hermsen et al.
Toxicology in vitro : an international journal published in association with BIBRA, 25(3), 745-753 (2011-01-18)
The zebrafish embryotoxicity test (ZET) is a fast and simple method to study chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis in ovo. We developed a novel quantitative evaluation method to assess the development of the zebrafish
Rui-Sheng Wang et al.
Industrial health, 45(4), 574-578 (2007-09-20)
Ethylene glycol monoethyl ether (EGEE) can cause damage to testes and sperm, and its metabolites are believed to play an important role in its toxicity. Aldehyde dehydrogenase 2 (ALDH2) is involved in the metabolism of this chemical. To investigate whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service