Skip to Content
Merck
All Photos(1)

Documents

568554

Sigma-Aldrich

Tri(ethylene glycol) monoethyl ether

technical grade

Synonym(s):

Triethylene glycol monoethyl ether, Ethyltriglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2(OCH2CH2)3OH
CAS Number:
Molecular Weight:
178.23
Beilstein:
1700466
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

refractive index

n20/D 1.438 (lit.)

bp

256 °C (lit.)

density

1.020 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCCOCCO

InChI

1S/C8H18O4/c1-2-10-5-6-12-8-7-11-4-3-9/h9H,2-8H2,1H3

InChI key

WFSMVVDJSNMRAR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabella Orienti et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 54(2), 229-233 (2002-08-23)
Among the different methods used to increase the aqueous drug solubility, the preparation of a solid dispersion with a soluble carrier represents an interesting formulative approach. We substituted polyvinylalcohol with triethyleneglycolmonoethylether and obtained a suitable material for the formulation of
I Orienti et al.
Biomacromolecules, 6(5), 2875-2880 (2005-09-13)
A series of poly(vinyl alcohol) amphiphilic derivatives have been prepared to obtain polymeric aggregates in aqueous phase holding thermodynamic instability. The aim was to evaluate their ability to interact with tumor cells eliciting selective cytotoxicity. The poly(vinyl alcohol) derivatives were
Marco Lucarini et al.
Journal of the American Chemical Society, 126(30), 9326-9329 (2004-07-30)
EPR spectroscopy has been used to study the interaction of para-substituted benzyl hydroxyalkyl nitroxides with the monolayer of water-soluble protected gold cluster made by a short alkyl chain and a triethylene glycol monomethyl ether unit. The inclusion of nitroxide probes
[Establishment of the maximum permissible concentration triethylene glycol ethyl ether in reservoir water].
V A Kondratiuk et al.
Gigiena i sanitariia, (5)(5), 84-85 (1982-05-01)
Magnus Hummelgård et al.
PloS one, 6(2), e17209-e17209 (2011-03-11)
Metallic nanoparticle inks are used for printed electronics, but to reach acceptable conductivity the structures need to be sintered, usually using a furnace. Recently, sintering by direct resistive heating has been demonstrated. For a microscopic understanding of this Joule heating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service