Skip to Content
Merck
All Photos(1)

Key Documents

769495

Sigma-Aldrich

Cobalt(II) chloride hexahydrate

≥97%

Synonym(s):

Cobaltous chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

40 mmHg ( 0 °C)

Quality Level

Assay

≥97%
97.0-102.0% (KT)

form

solid

anion traces

nitrate (NO3-): ≤0.01%
sulfate (SO42-): ≤0.007%

cation traces

Fe: ≤0.005%
Ni: ≤0.15%
Pb: ≤0.002%
Zn: ≤0.05%

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II) chloride hexahydrate, a hydrated form of cobalt chloride, is employed in electroplating and catalyst preparation. It serves as a precursor for synthesizing electrode materials for lithium-ion batteries and acts as a catalyst in a range of organic reactions, including acetylation, tosylation of alcohols, and condensation reactions.

Application

Cobalt(II) chloride hexahydrate can be used as:      
  • An additive to the electron transport layer (ETL) in perovskite solar cells to improve their performance, particularly by reducing energy losses and increasing the open-circuit voltage.      
  • A cobalt source for doping ZnO nanostructures. The incorporation of cobalt ions into the ZnO matrix is crucial for modifying its electronic and optical properties.      
  • A precursor to modify cobalt metal-organic framework (Co-MOF) derived carbon microspheres for application as anode materials in lithium-ion batteries.

Analysis Note

Substances not precipitated by ammonium sulfide (as sulphates) ≤ 0.3 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step synthesis of cobalt and nitrogen co-doped carbon nanotubes and their catalytic activity for the oxygen reduction reaction
Fu, S., et al.
Journal of Material Chemistry A, 3, 12718-12722 (2015)
Hyeohn Kim et al.
ACS nano, 15(1), 979-988 (2020-12-18)
Chiral inorganic nanomaterials have revealed opportunities in various fields owing to their strong light-matter interactions. In particular, chiral metal oxide nanomaterials that can control light and biochemical reactions have been highlighted due to their catalytic activity and biocompatibility. In this
Electroplating and characterization of cobalt-nickel-iron and nickel-iron for magnetic microsystems applications
Rasmussen, FE., et al.
Sensors and actuators A, Physical, 92, 242-248 (2001)
Sebastian Klemenz et al.
ChemSusChem, 11(18), 3150-3156 (2018-07-27)
High-performance catalysts for the oxygen-evolution reaction in water electrolysis are usually based on expensive and rare elements. Herein, mixed-metal borides are shown to be competitive with established electrocatalysts like noble metal oxides and other transition-metal(oxide)-based catalysts. Iron incorporation into nanoscale
Excellent lithium ion storage property of porous MnCo2O4 nanorods
Zeng, P., et al.
Royal Society of Chemistry Advances, 6, 23074-23084 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service