Skip to Content
Merck
All Photos(1)

Key Documents

251755

Sigma-Aldrich

Osmium tetroxide solution

4 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OsO4
CAS Number:
Molecular Weight:
254.23
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
liquid

grade

for analytical purposes

vapor pressure

1.23 psi ( 20 °C)
3.55 psi ( 55 °C)

form

liquid

reaction suitability

reagent type: oxidant

concentration

3.5-4.5% (OsO4)
4 wt. % in H2O

density

1.0 g/mL at 20 °C
1.04 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

O=[Os](=O)(=O)=O

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

OsO4 (4wt% in H2O) is widely employed in organic synthesis especially for hydroxylation reactions. Additionally, it is also involved in the oxidative cleavage of alkenes.

Application


  • In situ X-ray-assisted electron microscopy staining for large biological samples.: This study explores the use of osmium tetroxide solution for X-ray-assisted electron microscopy staining, enhancing the visualization of large biological samples (Ströh et al., 2022).

  • Application of the electron microscope to the cytochemical peroxidase reaction in salamander leukocytes.: The use of osmium tetroxide solution in electron microscopy enhances the cytochemical analysis of peroxidase reactions in salamander leukocytes, providing clear and detailed images (MITSUI, 1960).


Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric induction in the reaction of osmium tetroxide with olefins.
Hentges SG and Barry Sharpless K.
Journal of the American Chemical Society, 102(12), 4263-4265 (1980)
On stereochemistry of osmium tetroxide oxidation of allylic alcohol systems: empirical rule.
Cha JK, et al.
Tetrahedron Letters, 24(37), 3943-3946 (1983)
Kinetics of Oxidation of Some Fluoroquinolones by Hexacyanoferrate (III) in Alkaline Medium.
Diab N, et al.
International Journal of Chemistry (Canadian Center of Science and Education), 34(2), 1388-1388 (2013)
Manuel Marx et al.
Nature protocols, 7(2), 394-407 (2012-02-04)
In this report, we describe a reliable protocol for biocytin labeling of neuronal tissue and diaminobenzidine (DAB)-based processing of brain slices. We describe how to embed tissues in different media and how to subsequently histochemically label the tissues for light
Yukihiko Sugita et al.
The Journal of general virology, 92(Pt 11), 2485-2493 (2011-07-29)
Negatively stained influenza virions sometimes show irregular morphology and are often referred to as pleomorphic. However, this irregular morphology has not been visualized when ultrathin-section transmission and scanning electron microscopies are used. This study focused on the effects of ultracentrifugation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service