Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
OsO4
CAS Number:
Molecular Weight:
254.23
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
MDL number:
Form:
solution
form
solution
Quality Segment
reaction suitability
reagent type: oxidant
concentration
4% in H2O
technique(s)
electron microscopy: suitable
color
colorless to yellow
pH
6.0-8.0
density
1.0 g/mL at 20 °C
application(s)
hematology
histology
storage temp.
2-8°C
SMILES string
O=[Os](=O)(=O)=O
InChI
1S/4O.Os
InChI key
VUVGYHUDAICLFK-UHFFFAOYSA-N
Application
Osmium tetroxide solution has been used as a fixative in scanning and transmission electron microscopy.
Biochem/physiol Actions
Osmium tetroxide is also referred to as osmium and is mainly used as a secondary fixative of tissues. The molecule reacts with lipid moieties and is responsible for the oxidation of unsaturated bonds of fatty acids. This reaction adds density and contrast to biological samples. In normal tissues, presence of osmium results in intense black staining which can be viewed by normal eye. In addition, it can be used as a mordant (a molecule which combines with stain or dye at later stages) in electron microscopy, thereby enhancing lead staining.
Packaging
5 mL in ampule
10 mL in ampule
10 mL in ampule
Other Notes
Due to the hazardous nature of osmium tetroxide, it is recommended to use an ampule opener when using this chemical.
Still not finding the right product?
Explore all of our products under Osmium tetroxide solution
signalword
Danger
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

