Skip to Content
Merck
All Photos(6)

Key Documents

190500

Sigma-Aldrich

tert-Butyldimethylsilyl chloride

reagent grade, 97%

Synonym(s):

tert-Butyl(chloro)dimethylsilane, tert-Butyldimethylchlorosilane, TBDMSCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2Cl
CAS Number:
Molecular Weight:
150.72
Beilstein:
505999
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

Assay

97%

form

solid

bp

125 °C (lit.)

mp

86-89 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)Cl

InChI

1S/C6H15ClSi/c1-6(2,3)8(4,5)7/h1-5H3

InChI key

BCNZYOJHNLTNEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

tert-Butyldimethylsilyl chloride (TBDMSCl) is an organosilicon compound that can be used as a versatile protecting reagent for alcohols, amines, amides, and various carboxylic acids. It is also used in the preparation of isoxazoline N-oxides from α-bromonitroalkanes and olefins. TBDMSCl is used as a precursor that forms a silicon substrate for Ag/ZnO/graphene-based nanocomposites to form effective photocatalytic system for hydrogen production.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1A

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effective silylation of carboxylic acids under solvent-free conditions with tert-butyldimethylsilyl chloride (TBDMSCL) and triisopropylsilyl chloride (TIPSCL)
Firouzabadi H, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 166(1) (2000)
Ag/ZnO/graphene-tert-butyldimethylsilyl chloride hybrid nanocomposite as highly efficient catalyst for hydrogen production
Attia YA
Materials Express, 6(3) (2016)
Journal of the Chemical Society. Chemical Communications, 125-125 (1993)
Greene, T.W. Wuts, P.G.M.
Protective Groups in Organic Synthesis (1991)
New approach for the synthesis of isoxazoline-N-oxides
Kunetsky RA, et al.
Organic Letters, 5(25) (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service