Skip to Content
Merck
All Photos(1)

Documents

A65603

Sigma-Aldrich

4-(Aminomethyl)pyridine

98%

Synonym(s):

4-Picolylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
108057
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

230 °C (lit.)

mp

−8 °C (lit.)

density

1.065 g/mL at 25 °C (lit.)

SMILES string

NCc1ccncc1

InChI

1S/C6H8N2/c7-5-6-1-3-8-4-2-6/h1-4H,5,7H2

InChI key

TXQWFIVRZNOPCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[Synthesis of 3-nitro-1-nitroso-1-(4-pyridylmethyl)guanidine and related compounds].
S Kamiya
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (102)(102), 105-108 (1984-01-01)
Yuguang Ma et al.
The journal of physical chemistry. B, 111(10), 2711-2718 (2007-02-24)
HO-(TPA)FeV=O (TPA = tris(2-pyridylmethyl)amine) has been proposed in the literature as the key high-valent iron-oxo intermediate involved in alkane hydroxylation. Here the structure of this species is investigated theoretically in the framework of density functional theory (DFT). A detailed electronic
Sudhakar Ganta et al.
Inorganic chemistry, 57(9), 5145-5158 (2018-04-25)
A series of self-assembled binuclear complexes (1-4) of Pd2L'2L2 composition are prepared by combination of selected cis-protected palladium(II) component cis-PdL' with equimolar amount of the nonchelating bidentate ligand 1,3-bis((pyridine-4-yl)methyl)urea, L. The cis-protecting unit L' crafted in the complexes [Pd2(L')2(L)2](NO3)4, 1-4
Datong Wu et al.
The Analyst, 145(9), 3395-3400 (2020-04-03)
Here, ionized chiral carbon dots, (S,S)-C-dots-1 (λex = 430 nm, λem = 480 nm), were synthesized via a facile route with relatively high quantum yield (∼24.4%) and used as a fluorescent chiral sensor. One of the advantages of the synthetic
Meenakshi Verma et al.
Journal of materials chemistry. B, 8(36), 8405-8413 (2020-09-24)
Okadaic acid (OA) is one of the known marine biotoxins produced by various dinoflagellates and exists in seafood such as shellfish. The consumption of contaminated shellfish with OA leads to diarrheic shellfish poisoning (DSP), which results in the inhibition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service