Skip to Content
Merck
All Photos(4)

Key Documents

237086

Sigma-Aldrich

Aluminum potassium sulfate dodecahydrate

ACS reagent, ≥98%

Synonym(s):

Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlK(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
474.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

Assay

≥98%
98.0-102.0% (ACS specification)

form

powder or crystals

reaction suitability

reagent type: catalyst
core: aluminum

impurities

≤0.005% insolubles

pH

3.3 (94.88 g/L)

mp

92 °C (lit.)

density

1.757 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm

cation traces

Fe: ≤0.001%
NH4+: ≤0.005%
Na: ≤0.02%
heavy metals: ≤0.001% (by ICP)

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

GNHOJBNSNUXZQA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Aluminum potassium sulfate dodecahydrate has been used:
  • to prepare Mayer′s hematoxylin solution for staining of lung sections and tibialis anterior muscle sections
  • to sensitize experimental mice and establish the allergic rhinitis (AR) mouse model
  • to stain mammary glands for its whole-mount analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Wang et al.
Bio-protocol, 7(10) (2017-07-29)
Muscle function is determined by its structure and fiber type composition. Here we describe a protocol to examine muscle histology and myofiber types using hematoxylin and eosin (H&E) and immunofluorescence staining, respectively. H&E stain nucleus in blue and cytoplasm in
Michael S Sinyakov et al.
Vaccine, 24(42-43), 6534-6541 (2006-07-18)
Bovine serum albumin (BSA) and the surface A-layer protein (AP) of an atypical strain of fish bacterial pathogen Aeromonas salmonicida were covalently linked with polymeric nano- and microparticles, and antigenicity of the resulted conjugates was compared in mice and goldfish.
Growth rates of aluminum potassium sulfate crystals in aqueous solutions.
Botsaris GD and Denk Jr EG.
Industrial & Engineering Chemistry Fundamentals, 9(2), 276-283 (1970)
Saraswoti Khadge et al.
Journal of mammary gland biology and neoplasia, 23(1-2), 43-58 (2018-03-27)
Studies in rodents have shown that dietary modifications as mammary glands (MG) develop, regulates susceptibility to mammary tumor initiation. However, the effects of dietary PUFA composition on MGs in adult life, remains poorly understood. This study investigated morphological alterations and
Eagleson M.
Concise Encyclopedia Chemistry, 886-886 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service