Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum potassium sulfate dodecahydrate
Synonym(s):
Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate
Linear Formula:
AlK(SO
4
)
2
· 12H
2
O
CAS Number:
7784-24-9
Molecular Weight:
474.39
EC Number:
233-141-3
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (4)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
237086
Aluminum potassium sulfate dodecahydrate,
ACS reagent, ≥98%
Expand
View Pricing
31242
Aluminum potassium sulfate dodecahydrate,
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Expand
View Pricing
A6435
Aluminum potassium sulfate dodecahydrate,
BioReagent, suitable for insect cell culture
Expand
View Pricing
A7210
Aluminum potassium sulfate dodecahydrate,
BioXtra, ≥98.0%
Expand
View Pricing