All Photos(2)
About This Item
Empirical Formula (Hill Notation):
AgBr
CAS Number:
Molecular Weight:
187.77
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
form:
powder, crystals or chunks
Recommended Products
Quality Level
Assay
99%
form
powder, crystals or chunks
mp
432 °C (lit.)
density
6.473 g/mL at 25 °C (lit.)
SMILES string
Br[Ag]
InChI
1S/Ag.BrH/h;1H/q+1;/p-1
InChI key
ADZWSOLPGZMUMY-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ji Hye Seo et al.
Journal of hazardous materials, 198, 347-355 (2011-11-18)
Photocytotoxicity of visible-light catalytic Ag/AgBr/TiO(2) nanoparticles (NPs) was examined both in vitro and in vivo. The Ag/AgBr/TiO(2) NPs were prepared by the deposition-precipitation method. Their crystalline structures, atomic compositions, and light absorption property were examined by X-ray diffraction (XRD) patterns
Wenxin Wang et al.
Journal of hazardous materials, 243, 169-178 (2012-11-06)
A simple microemulsion-like chemical precipitation method has been successfully developed to construct effectively-contacted AgBr-TiO(2) composite. The key of this method is the dual roles of Br(-) in the synthetic process, as linkers between cetyltrimethyl ammonium cation surfactants and nanocrystalline anatase
The theory of the photolysis of silver bromide and the photographic latent image.
Gurney RW and Mott NF.
Proceedings of the Royal Society of London A: Mathematical, Physical and Engineering Sciences., 151-167 (1938)
Fabien Hubert et al.
Langmuir : the ACS journal of surfaces and colloids, 24(17), 9219-9222 (2008-08-12)
A complex between cetyltrimethylammonium bromide (CTAB) surfactant and silver bromide (CTASB) is recognized by NMR and X-ray photoelectron spectroscopy (XPS) to be the entity at the surface of gold nanorods, resulting from an in situ formation in the classical scheme
Lattice defects in silver bromide.
Berry CR.
Physical Review, 82(3), 422-422 (1951)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service