Skip to Content
Merck

Skip To

85193

Silver nitrate solution

2.5 % (w/v) AgNO3 in H2O

Synonym(s):

Argentum nitricum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
MDL number:
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

concentration

2.5 % (w/v) AgNO3 in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

General description

Silver nitrate along with Iodine are among several inorganic substances used to induce color in microscopical preparations. Silver nitrate as a biological stain exploits the strong colors of colloidal metallic silver. Silver nitrate is an important component of the silver staining that is used for the detection of proteins separated by polyacrylamide gel electrophoresis.

Application

Silver nitrate is a component of the Silver stain kit. It is required for image development in diamine as well as non-diamine silver staining.

Biochem/physiol Actions

In silver staining, silver ions are reduced to metallic silver, thereby giving metallic silver images.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.090811.090801.09990
form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

-

application(s)

-

application(s)

-

concentration

2.5 % (w/v) AgNO3 in H2O

concentration

0.1 M

concentration

1 M

concentration

0.1 M

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C


Still not finding the right product?

Explore all of our products under Silver nitrate solution


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters