Skip to Content
Merck
All Photos(1)

Documents

02151

Sigma-Aldrich

Sodium bromate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBrO3
CAS Number:
Molecular Weight:
150.89
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38050507
PubChem Substance ID:
NACRES:
NA.21

Agency

suitable for EPA 300

Assay

≥99%

form

crystals

impurities

≤0.005% heavy metals (as Pb)

pH

5-9 (25 °C, 50 g/L)

density

3.339 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤500 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

[Na+].[O-]Br(=O)=O

InChI

1S/BrHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

XUXNAKZDHHEHPC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bromate is a commercially available, very stable oxidant. It is much easier to handle than liquid bromine or hypobromous acid solutions. The elastic, piezoelectric and dielectric constants of sodium chlorate and sodium bromate (NaBrO3) have been evaluated.

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Ox. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium-catalyzed oxidation of alcohols with sodium bromate.
Yamamoto Y, et al.
Tetrahedron Letters, 26(17), 2107-2108 (1985)
Autocatalytic oxidation of ethers with sodium bromate.
Metsger L and Bittner S.
Tetrahedron, 56(13), 1905-1910 (2000)
Elastic, Piezoelectric, and Dielectric Properties of Sodium Chlorate and Sodium Bromate.
Mason WP.
Physical Review, 70(7-8), Elastic-Elastic (1946)
An improved method for the synthesis of ?-lactones using sodium bromate and sodium hydrogen sulfite.
Hayat S, et al.
Tetrahedron Letters, 42(9), 1647-1649 (2001)
Cerium or ruthenium catalyzed oxidation of alcohols to carbonyl compounds by means of sodium bromate.
Kanemoto S, et al.
Bulletin of the Chemical Society of Japan, 59(1), 105-108 (1985)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service