Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium bromate
Synonym(s):
Sodium bromate
Linear Formula:
NaBrO
3
CAS Number:
7789-38-0
Molecular Weight:
150.89
EC Number:
232-160-4
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
pH Value
Physical Form
Purity
Agency
Quality Segment
Reagent Type
Special Grade
Puriss (2)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
02151
Sodium bromate,
≥99%
Expand
View Pricing
8.14368
Sodium bromate,
for synthesis
Expand
View Pricing
71325
Sodium bromate,
puriss. p.a., ≥99.5% (RT)
Expand
View Pricing