Skip to Content
Merck
All Photos(2)

Key Documents

213462

Sigma-Aldrich

Sodium borohydride

ReagentPlus®, 99%

Synonym(s):

Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22
grade:
reagent
form:
powder

grade

reagent

product line

ReagentPlus®

Assay

99%

form

powder

reaction suitability

reagent type: reductant

mp

>300 °C (dec.) (lit.)

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium borohydride, also known as sodium tetrahydridoborate, is a highly basic boron hydride that is commonly used as a reducing agent. It reacts very slowly with water and acts as a catalyst and stabilizer.

Application

Sodium borohydride can be used as a reducing agent for the:
  • Synthesis of cobalt (II, III) oxide (Co3O4) from cobalt chloride (CoCl2) via a co-reduction process.
  • Solvent-free reduction of aldehydes and ketones to alcohols in the presence of solid acid as an activator.
  • Reductive amination of carbonyl compounds to secondary amines in the presence of silica-gel-supported sulfuric acid as a catalyst.
  • Raney nickel-catalyzed reduction of aromatic nitro compounds to aromatic amines.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic Co 3 O 4/reduced graphene oxide nanocomposite as a superior heterogeneous catalyst for one-pot oxidative esterification of aldehydes to methyl esters
Vineeta P,et al.
Royal Society of Chemistry Advances, 5, 88567-88573 (2015)
Sodium borohydride versus ammonia borane, in hydrogen storage and direct fuel cell applications
Umit DB,et al.
Energy & Environmental Science, 2, 627-637 (2009)
Kinetics of Ru-catalyzed sodium borohydride hydrolysis
JS Z,et al.
Journal of Power Sources, 164, 772-781 (2007)
Dinesh Bhalothia et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-07-25)
Herein, ternary metallic nanocatalysts (NCs) consisting of Au clusters decorated with a Pt shell and a Ni oxide core underneath (called NPA) on carbon nanotube (CNT) support were synthesized by combining adsorption, precipitation, and chemical reduction methods. By a retrospective
Nicholas A Harris et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(42), 8922-8942 (2018-08-29)
Stress is a precipitating agent in neuropsychiatric disease and initiates relapse to drug-seeking behavior in addicted patients. Targeting the stress system in protracted abstinence from drugs of abuse with anxiolytics may be an effective treatment modality for substance use disorders.

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service