Skip to Content
Merck
All Photos(1)

Key Documents

Y0001064

Benzaldehyde

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Bitter almond

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHO
CAS Number:
Molecular Weight:
106.12
Beilstein:
471223
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

3.7 (vs air)

vapor pressure

4 mmHg ( 45 °C)

API family

benzaldehyde

autoignition temp.

374 °F

expl. lim.

1.4 %, 20 °F

manufacturer/tradename

EDQM

refractive index

n20/D 1.545 (lit.)

bp

178-179 °C (lit.)

mp

−26 °C (lit.)

density

1.044 g/cm3 at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=Cc1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H

InChI key

HUMNYLRZRPPJDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzaldehyde EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-gen Peng et al.
Chemical communications (Cambridge, England), 49(26), 2709-2711 (2013-02-26)
A center radially fibrous silica encapsulated TS-1 zeolite (TS-1@KCC-1) has been synthesized in a microemulsion system for the first time. Supporting the Rh(OH)3 species, this novel core-shell structured material serves as a robust bifunctional catalyst for one-pot synthesis of benzamide
Longyun Lv et al.
ACS combinatorial science, 15(4), 183-192 (2013-02-22)
We previously reported the novel efficient proton/heat-promoted four-component reactions (4CRs) of but-2-ynedioates, two same/different primary amines, and aldehydes for the synthesis of tetra- and pentasubstituted polyfunctional dihydropyrroles. If aromatic and aliphatic amines were used as reagents, four different series of
Lidia Mateo et al.
PLoS genetics, 10(8), e1004560-e1004560 (2014-08-15)
The increase in availability of whole genome sequences makes it possible to search for evidence of adaptation at an unprecedented scale. Despite recent progress, our understanding of the adaptive process is still very limited due to the difficulties in linking
Haiqiang Jin et al.
International journal of pharmaceutics, 434(1-2), 384-390 (2012-06-12)
To find a way to modulate the effect of thrombolytic proteins by increasing their specificity, minimizing their adverse effect as well as lengthening their circulation time for the treatment of ischemic vascular disease holds great promise. In this work, urokinase-type
Thermoresponsive dynamic covalent single-chain polymer nanoparticles reversibly transform into a hydrogel.
Daniel E Whitaker et al.
Angewandte Chemie (International ed. in English), 52(3), 956-959 (2012-12-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service