Skip to Content
Merck
All Photos(4)

Key Documents

493511

Sigma-Aldrich

Ethyl alcohol, Pure

greener alternative

≥95.0%, ACS spectrophotometric grade, 190 proof, suitable for UV/Vis spectroscopy

Synonym(s):

Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS spectrophotometric grade
technique(s):
UV/Vis spectroscopy: suitable
bp:
78.3 °C
Pricing and availability is not currently available.

Product Name

Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%

grade

ACS spectrophotometric grade

Quality Level

vapor density

1.59 (vs air)

Assay

95.0%

form

liquid

expl. lim.

3.1-27.7 %

availability

not available in (Sales restictions may apply)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

190 proof

technique(s)

UV/Vis spectroscopy: suitable

impurities

H2SO4, passes test (darkened)
MnO4- reducers, passes test
acetone, passes test
isopropanol, passes test
≤0.0002 meq/g Titr. base
≤0.0005 meq/g Titr. acid
≤0.1% methanol

evapn. residue

≤0.001%

color

APHA: ≤10

bp

78.3 °C

mp

-114 °C

solubility

H2O: passes test

density

0.816 g/mL at 25 °C

λ

H2O reference

UV absorption

λ: 210 nm Amax: 0.40
λ: 220 nm Amax: 0.25
λ: 230 nm Amax: 0.15
λ: 240 nm Amax: 0.05
λ: 270-400 nm Amax: 0.01

greener alternative category

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prices include Federal Tax.
Pure ethyl alcohol is a non-denatured 190proof ethanol. Ethanol, a primary alcohol, is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide.[1] The impact of ethanol/water fumigation on the performance of diesel engines has been studied.[2] Spectrophotometric grade solvent are of high purity grade with low UV absorption.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". 

Application


  • Monitoring and evaluation of alcoholic fermentation processes using a chemocapacitor sensor array.: Ethyl alcohol is studied not just as a product but also as an analytical tool in monitoring fermentation processes. This work illustrates the integration of sensor technology with chemical analysis, highlighting advancements in analytical techniques for monitoring and quality control in the beverage industry (Oikonomou et al., 2014).

  • Ancient nigella seeds from Boyali Höyük in north-central Turkey.: Utilizing ethyl alcohol in the conservation and analysis of ancient botanical remains, this historical study reflects the critical role of analytical chemistry in archaeological research, providing insights into ancient agricultural practices (Salih et al., 2009).

Other Notes

Sales restrictions may apply

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of a direct injection diesel engine fumigated with ethanol/water mixtures.
Morsy MH.
Energy Conversion and Management , 94, 406-414 (2015)
Sodium hydroxide-catalyzed transfer hydrogenation of carbonyl compounds and nitroarenes using ethanol or isopropanol as both solvent and hydrogen donor.
Wang D, et al.
J. Mol. Catal. A: Chem., 400, 14-21 (2015)
N Di Girolamo et al.
Stem cells (Dayton, Ohio), 33(1), 157-169 (2014-06-27)
Stem cell (SC) division, deployment, and differentiation are processes that contribute to corneal epithelial renewal. Until now studying the destiny of these cells in a living mammal has not been possible. However, the advent of inducible multicolor genetic tagging and
Ioannis Lignos et al.
Small (Weinheim an der Bergstrasse, Germany), 11(32), 4009-4017 (2015-05-23)
The early-time kinetics (<1 s) of lead sulfide (PbS) quantum dot formation are probed using a novel droplet-based microfluidic platform, which allows for high-throughput and real-time optical analysis of the reactive process with millisecond time resolution. The reaction platform enables
Ross B Mounsey et al.
Experimental neurology, 273, 36-44 (2015-08-06)
Parkinson's disease (PD) is a common chronic neurodegenerative disorder, usually of idiopathic origin. Symptoms including tremor, bradykinesia, rigidity and postural instability are caused by the progressive loss of dopaminergic neurons in the nigrostriatal region of the brain. Symptomatic therapies are

Articles

This application note describes the HPTLC-based identification of fixed oils using octadecyl silyl silica gel as the coating substance, following USP 43 NF 38 monograph method containing USP general chapter <202>.

This application note describes the HPTLC-based identification of fixed oils using octadecyl silyl silica gel as the coating substance, following USP 43 NF 38 monograph method containing USP general chapter <202>.

This application note describes the HPTLC-based identification of fixed oils using octadecyl silyl silica gel as the coating substance, following USP 43 NF 38 monograph method containing USP general chapter <202>.

This application note describes the HPTLC-based identification of fixed oils using octadecyl silyl silica gel as the coating substance, following USP 43 NF 38 monograph method containing USP general chapter <202>.

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is there denatured ethanol available that is certified as Kosher, FCC, and non-GMO?

    1 answer
    1. No, there is no denatured ethanol available that is certified as Kosher, FCC, and non-GMO

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service