Skip to Content
Merck
All Photos(1)

Key Documents

1050993

USP

Benzalkonium Chloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Alkylbenzyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benzalkonium chloride

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Benzalkonium Chloride is a mixture of alkylbenzyldimethylammonium chlorides.

Application

Benzalkonium Chloride USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, system suitability, and identification solution during the assay, and impurity analysis by liquid chromatography in conjunction with a UV detector according to the given below monographs of United States Pharmacopeia (USP):
  • Benzalkonium Chloride
  • Benzalkonium Chloride Solution
  • Fluticasone Propionate Nasal Spray
  • Triamcinolone Acetonide Nasal Spray

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzalkonium Chloride
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 38(2), 5634-5634 (2013)
Fluticasone Propionate Nasal Spray
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(6), 2000-2000 (2023)
Benzalkonium Chloride Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(5), 5636-5636 (2016)
Triamcinolone Acetonide Nasal Spray
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(5) (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service