Skip to Content
Merck
All Photos(1)

Key Documents

Y0000684

Trimethoprim for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Trimethoprim, 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, NSC 106568

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
Beilstein:
625127
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trimethoprim

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC

InChI

1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)

InChI key

IEDVJHCEMCRBQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Primarily used as an antibacterial agent. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai Liu et al.
Journal of hazardous materials, 235-236, 367-375 (2012-08-25)
The preparation of activated carbons (AC-H(x)P(y)O(z)) by four kinds of oxyacids of phosphorus (H(3)PO(4), H(4)P(2)O(7), HPO(3) and H(3)PO(3)) activation of lotus stalk (LS) was studied, with a particular focus on the effect of these H(x)P(y)O(z) on both surface chemistry and
Kyuho Han et al.
Nature methods, 11(1), 86-93 (2013-11-12)
Protein concentrations are often regulated by dynamic changes in translation rates. Nevertheless, it has been challenging to directly monitor changes in translation in living cells. We have developed a reporter system to measure real-time changes of translation rates in human
J T Andersen et al.
Epidemiology and infection, 141(8), 1749-1755 (2012-09-27)
The antibiotic trimethoprim acts as a folate antagonist. Since trophoblasts are very sensitive to drugs that interfere with the folic acid cycle and thereby inhibit DNA synthesis, use of trimethoprim during the first trimester could be associated with miscarriage. A
Khalid Tai et al.
PloS one, 7(9), e46269-e46269 (2012-10-03)
Regulating transgene expression in vivo by delivering oral drugs has been a long-time goal for the gene therapy field. A novel gene regulating system based on targeted proteasomal degradation has been recently developed. The system is based on a destabilizing
Ivan Senta et al.
Water research, 47(2), 705-714 (2012-11-29)
Comprehensive study on the occurrence and fate of several classes of antimicrobials, including sulfonamides, trimethoprim, fluoroquinolones and macrolides, in Croatian municipal wastewaters was performed using an integrated approach, which comprised analysis of both dissolved and particulate fractions. A nation-wide screening

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service