Skip to Content
Merck
All Photos(1)

Key Documents

S2100000

Sulfamethoxazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Amino-N-(5-methyl-3-isoxazolyl)benzenesulfonamide, N1-(5-Methylisoxazol-3-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N3O3S
CAS Number:
Molecular Weight:
253.28
Beilstein:
6732984
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfamethoxazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(NS(=O)(=O)c2ccc(N)cc2)no1

InChI

1S/C10H11N3O3S/c1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h2-6H,11H2,1H3,(H,12,13)

InChI key

JLKIGFTWXXRPMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfamethoxazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Trimethoprim-sulfamethoxazole-associated aseptic meningitis: case reports and review of the literature.
A M Joffe et al.
The American journal of medicine, 87(3), 332-338 (1989-09-01)
Alejandro Hoberman et al.
The New England journal of medicine, 370(25), 2367-2376 (2014-05-06)
Children with febrile urinary tract infection commonly have vesicoureteral reflux. Because trial results have been limited and inconsistent, the use of antimicrobial prophylaxis to prevent recurrences in children with reflux remains controversial. In this 2-year, multisite, randomized, placebo-controlled trial involving
Co-trimoxazole (trimethoprim-sulfamethoxazole): an updated review of its antibacterial activity and clinical efficacy.
G P Wormser et al.
Drugs, 24(6), 459-518 (1982-12-01)
D H Lawson et al.
Reviews of infectious diseases, 4(2), 429-433 (1982-03-01)
A comprehensive review is presented of the world literature on adverse reactions to trimethoprim-sulfamethoxazole (TMP-SMZ) since its first use in 1968, when concern was expressed about possible hematologic toxicity. Serious toxicity is a rare event, but when it occurs severe
Jih-Tay Hsu et al.
Journal of hazardous materials, 277, 34-43 (2014-03-19)
Antibiotics are commonly used in swine feed to treat and prevent disease, as well as to promote growth. Antibiotics released into the environment via wastewater could accelerate the emergence of antibiotic-resistant bacteria and resistance genes in the surrounding environment. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service