Skip to Content
Merck
All Photos(1)

Key Documents

Y0000420

Cefotaxime acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cefotaxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N5O7S2
CAS Number:
Molecular Weight:
455.47
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefotaxime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[s]1c(nc(c1)\C(=N\OC)\C(=N\[C@@H]2[C@@H]3SCC(=C(N3C2=O)C(=O)O)COC(=O)C)\[O-])N

InChI

1S/C16H17N5O7S2.Na/c1-6(22)28-3-7-4-29-14-10(13(24)21(14)11(7)15(25)26)19-12(23)9(20-27-2)8-5-30-16(17)18-8;/h5,10,14H,3-4H2,1-2H3,(H2,17,18)(H,19,23)(H,25,26);/q;+1/p-1/b20-9-;/t10-,14-;/m0./s1

InChI key

AZZMGZXNTDTSME-XJTDMATHSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefotaxime acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Delangle et al.
Medecine et maladies infectieuses, 43(6), 244-247 (2013-06-29)
We had for aim to check the appropriateness of our practices according to French guidelines (17th consensus conference, SPILF 2008) and to identify variables associated with the delay before appropriate measures were implemented. Our retrospective observational study (2009-2011) focused on
Elad Firnberg et al.
Nucleic acids research, 41(15), 7420-7428 (2013-06-12)
An important goal of evolutionary biology is to understand the constraints that shape the dynamics and outcomes of evolution. Here, we address the extent to which the structure of the standard genetic code constrains evolution by analyzing adaptive mutations of
Martijn F Schenk et al.
Molecular biology and evolution, 30(8), 1779-1787 (2013-05-17)
Understanding epistasis is central to biology. For instance, epistatic interactions determine the topography of the fitness landscape and affect the dynamics and determinism of adaptation. However, few empirical data are available, and comparing results is complicated by confounding variation in
Christine Ferreiro et al.
European journal of pediatrics, 172(9), 1243-1248 (2013-05-17)
The treatment of complicated urinary tract infection in children is still a matter of debate. In our hospital, antimicrobial treatment is initiated intravenously, and the duration of this treatment is adapted according to the results of a Tc-99m dimercaptosuccinic acid
Kurinchi Selvan Gurusamy et al.
The Cochrane database of systematic reviews, 8(8), CD010268-CD010268 (2013-08-21)
Risk of methicillin-resistant Staphylococcus aureus (MRSA) infection after surgery is generally low, but affects up to 33% of patients after certain types of surgery. Postoperative MRSA infection can occur as surgical site infections (SSIs), chest infections, or bloodstream infections (bacteraemia).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service