Skip to Content
Merck

A6987

Ceftazidime hydrate

Synonym(s):

1-[[(6R,7R)-7-[[(2Z)-(2-amino-4-thiazolyl)[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridinum

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C22H22N6O7S2 · xH2O
CAS Number:
Molecular Weight:
546.58 (anhydrous basis)
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
51284108
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

does not contain sodium carbonate

Quality Level

assay

95.0-102.0% anhydrous basis

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(C)(O\N=C(/C(=O)N[C@H]1[C@H]2SCC(C[n+]3ccccc3)=C(N2C1=O)C([O-])=O)c4csc(N)n4)C(O)=O

InChI

1S/C22H22N6O7S2.5H2O/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27;;;;;/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34);5*1H2/b26-13-;;;;;/t14-,18-;;;;;/m1...../s1

InChI key

NMVPEQXCMGEDNH-TZVUEUGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Broad spectrum antibiotic; third generation cephalosporin

Application

Ceftazidime hydrate is used to study the effect of expression, binding and inhibition of penicillin-binding proteins especially PBP3 on bacterial cell wall mucopeptide synthesis. It is also used in bacterial susceptibility studies and to select mutant colonies of interest.
Used to study the effect of expression, binding and inhibition of penicillin-binding proteins especially PBP3 on bacterial cell wall mucopeptide synthesis.

Biochem/physiol Actions

Cephalosporins, such as ceftazidime , disrupt the synthesis of the peptidoglycan layer of bacterial cell walls.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neang S Ly et al.
The Journal of antimicrobial chemotherapy, 70(5), 1434-1442 (2015-02-26)
Colistin is an 'old' drug, which is being increasingly utilized due to limited therapeutic options. However, resistance emergence during monotherapy is concerning. Here, our objective was to optimize colistin combinations against Pseudomonas aeruginosa by profiling the time course of synergistic
David M P De Oliveira et al.
Science translational medicine, 12(570) (2020-11-20)
The emergence of polymyxin resistance in carbapenem-resistant and extended-spectrum β-lactamase (ESBL)-producing bacteria is a critical threat to human health, and alternative treatment strategies are urgently required. We investigated the ability of the hydroxyquinoline analog ionophore PBT2 to restore antibiotic sensitivity
Borys Snopok et al.
The Analyst, 140(9), 3225-3232 (2015-03-13)
An ultra-sensitive gas phase biosensor/tracer/bio-sniffer is an emerging technology platform designed to provide real-time information on air-borne analytes, or those in liquids, through classical headspace analysis. The desired bio-sniffer measures gaseous 17α- ethinylestradiol (ETED) as frequency changes on a quartz
Ruud R G Bueters et al.
Pediatric research, 76(6), 508-514 (2014-08-29)
Many premature born neonates receive antibiotic drugs to treat infections, which are applied during active nephrogenesis. We studied the impact of clinical concentrations of gentamicin and alternatives, ceftazidime and meropenem, on ureteric branching. Mice metanephroi were dissected at embryonic day
Yung-Heng Hsu et al.
International journal of nanomedicine, 9, 4347-4355 (2014-09-24)
We developed biodegradable drug-eluting nanofiber-enveloped implants that provided sustained release of vancomycin and ceftazidime. To prepare the biodegradable nanofibrous membranes, poly(D,L)-lactide-co-glycolide and the antibiotics were first dissolved in 1,1,1,3,3,3-hexafluoro-2-propanol. They were electrospun into biodegradable drug-eluting membranes, which were then enveloped

Global Trade Item Number

SKUGTIN
A6987-5G04061833333471
A6987-1G04061833383155

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service