Skip to Content
Merck
All Photos(1)

Documents

Y0000199

Ciprofloxacin hydrochloride for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ciprofloxacin hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18FN3O3 · HCl · H2O
CAS Number:
Molecular Weight:
385.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ciprofloxacin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.Cl.OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O

InChI

1S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19H,1-6H2,(H,23,24);1H;1H2

InChI key

ARPUHYJMCVWYCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ciprofloxacin hydrochloride for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Chen et al.
Food chemistry, 148, 105-111 (2013-11-23)
Aqueous two-phase systems (ATPS) are potential industrial technology in separation and enrichment areas. However, the main problem is that ATPS could not be recycled, which results in a high cost and pollution of the environment. In this work, a new
Tong Yang et al.
Veterinary microbiology, 170(1-2), 89-96 (2014-02-21)
This study was performed to determine the prevalence of plasmid-mediated quinolone resistance (PMQR) determinants and characterize the ciprofloxacin resistance in Escherichia coli isolated from different sources in China. PMQR determinants were detected by PCR amplification and sequencing in 2297 E.
Risaku Fukumoto et al.
PloS one, 8(3), e58389-e58389 (2013-03-23)
Radiation combined injury (CI) is a radiation injury (RI) combined with other types of injury, which generally leads to greater mortality than RI alone. A spectrum of specific, time-dependent pathophysiological changes is associated with CI. Of these changes, the massive
Patrik Ellström et al.
International journal of medical microbiology : IJMM, 303(3), 134-139 (2013-03-27)
Recent studies have indicated a role of the lipooligosaccharide (LOS) of Campylobacter jejuni in the severe neurological Guillain Barré syndrome, as well as in development of more severe symptoms of acute enteritis. We evaluated the role of the LOS locus
Pieter Dewint et al.
Gut, 63(2), 292-299 (2013-03-26)
To assess whether a combination of adalimumab and ciprofloxacin is superior to adalimumab alone in the treatment of perianal fistulising Crohn's disease (CD). Randomised, double-blind, placebo controlled trial in eight Dutch hospitals. In total, 76 CD patients with active perianal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service