Skip to Content
Merck
All Photos(1)

Key Documents

1134335

USP

Ciprofloxacin hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ciprofloxacin hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18FN3O3 · HCl · H2O
CAS Number:
Molecular Weight:
385.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ciprofloxacin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.Cl.OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O

InChI

1S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19H,1-6H2,(H,23,24);1H;1H2

InChI key

ARPUHYJMCVWYCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucile Pantel et al.
Antimicrobial agents and chemotherapy (2021-03-10)
NOSO-502 is a preclinical antibiotic candidate of the Odilorhabdin class. This compound exhibits activity against Enterobacteriaceae pathogens, including carbapenemase-producing bacteria and most of the Colistin (CST)-resistant strains. Among a collection of CST-resistant Klebsiella pneumoniae strains harboring mutations on genes pmrAB
Andrea Meyerhoff et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 39(3), 303-308 (2004-08-13)
In August 2000, the US Food and Drug Administration (FDA) approved ciprofloxacin hydrochloride (Cipro; Bayer) for management of postexposure inhalational anthrax. This was the first antimicrobial drug approved by the FDA for use in treating an infection due to a
Corinne N Thompson et al.
Tropical medicine & international health : TM & IH, 19(10), 1284-1292 (2014-07-30)
The country of Fiji, with a population of approximately 870 000 people, faces a growing burden of several communicable diseases including the bacterial infection typhoid fever. Surveillance data suggest that typhoid has become increasingly common in rural areas of Fiji
Tarek Haddad et al.
Chemosphere, 115, 40-46 (2014-03-19)
The presence of pharmaceuticals, especially antibiotics, in the aquatic environment is of growing concern. Several studies have been carried out on the occurrence and environmental risk of these compounds. Ciprofloxacin (CIP), a broad-spectrum anti-microbial second-generation fluoroquinolone, is widely used in
Ting-Chi Yeh et al.
Cancer, 120(8), 1255-1262 (2014-01-15)
The purpose of the current study was to prevent bloodstream infection and invasive fungal infection (IFI) by administering prophylactic antibiotic and antifungal agents during intensive chemotherapy in patients being treated for acute leukemia. Prophylaxis treatment was administered during intensive chemotherapy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service