Skip to Content
Merck
All Photos(1)

Key Documents

Y0001394

Fusidic acid for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fusidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H48O6
CAS Number:
Molecular Weight:
516.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fusidic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[H][C@@]12CC[C@@]3(C)[C@@]([H])([C@H](O)C[C@@]4([H])\C([C@H](C[C@]34C)OC(C)=O)=C(/CC\C=C(\C)C)C(O)=O)[C@@]1(C)CC[C@@H](O)[C@H]2C

InChI

1S/C31H48O6/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36)/b26-20-/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-/m0/s1

InChI key

IECPWNUMDGFDKC-MZJAQBGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fusidic acid for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Suppresses nitric oxide lysis of pancreatic islet cells. Inhibits protein synthesis in prokaryotes by inhibiting the ribosome-dependent activity of G factor and translocation of peptidyl-tRNA.
Suppresses nitric oxide lysis of pancreatic islet cells; inhibits protein synthesis in prokaryotes.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin P Howden et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 42(3), 394-400 (2006-01-05)
Fusidic acid has activity against a range of pathogens but has mainly been used to treat staphylococcal infections. Fusidic acid monotherapy, especially topical preparations, has been strongly associated with the emergence of fusidic acid resistance among both methicillin-resistant Staphylococcus aureus
J Turnidge et al.
International journal of antimicrobial agents, 12 Suppl 2, S35-S44 (1999-10-21)
Resistance to fusidic acid is determined by a number of mechanisms. The best described are alterations in elongation factor G, which appear in natural mutants that are harboured at low rates in normal populations of staphylococci (10(6) to 10(8)). Altered
K Christiansen
International journal of antimicrobial agents, 12 Suppl 2, S3-S9 (1999-10-21)
Unlike trials conducted today on new antimicrobials, the introduction of fusidic acid was not accompanied by extensive studies on toxicity and side effects. The early studies on small numbers of patients reported fusidic acid to be a nontoxic drug with
J D Wilkinson
The British journal of dermatology, 139 Suppl 53, 37-40 (1999-02-17)
Fusidic acid is an antibiotic that belongs to a group of its own, the fusidanes. The molecule has a steroid-like structure but does not possess any steroid activity. The structure is thought to be responsible for the steroid-like high penetration
Helmut Schöfer et al.
European journal of dermatology : EJD, 20(1), 6-15 (2009-12-17)
Studies on the clinical efficacy of fusidic acid in skin and soft-tissue infections (SSTIs), notably those due to Staphylococcus aureus, are reviewed. Oral fusidic acid (tablets dosed at 250 mg twice daily, or a suspension for paediatric use at 20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service