Skip to Content
Merck
All Photos(1)

Key Documents

T2009000

Trilaurin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glyceryl tridodecanoate, 1,2,3-Tridodecanoylglycerol, 1,2,3-Trilauroylglycerol, Glycerol trilaurate, Glyceryl trilaurate, Tridodecanoin, Trilaurin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
CAS Number:
Molecular Weight:
639.00
Beilstein:
1730452
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trilaurin

manufacturer/tradename

EDQM

mp

46.5 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

InChI

1S/C39H74O6/c1-4-7-10-13-16-19-22-25-28-31-37(40)43-34-36(45-39(42)33-30-27-24-21-18-15-12-9-6-3)35-44-38(41)32-29-26-23-20-17-14-11-8-5-2/h36H,4-35H2,1-3H3

InChI key

VMPHSYLJUKZBJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trilaurin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Chen et al.
ACS applied materials & interfaces, 8(38), 25087-25095 (2016-09-15)
Photothermal therapy exerts its anticancer effect by converting laser radiation energy into hyperthermia using a suitable photosensitizer. This study reports development of nanostructured lipid carriers (NLCs) suitable for noninvasive oral delivery of a near-infrared photosensitizer dye IR780. The carrier encapsulating
Arie Gruzman et al.
Journal of cellular and molecular medicine, 16(3), 594-604 (2011-05-14)
We have recently generated lipophilic D-xylose derivatives that increase the rate of glucose uptake in cultured skeletal muscle cells in an AMP-activated protein kinase (AMPK)-dependent manner. The derivative 2,4:3,5-dibenzylidene-D-xylose-diethyl dithioacetal (EH-36) stimulated the rate of glucose transport by increasing the
Dong-Jin Jang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 80, 162-172 (2016-05-03)
This study aimed to enhance the in vivo antitumor effects of camptothecin (CPT), a strong antitumor agent whose delivery is limited by poor aqueous solubility and instability of the active lactone form. CPT was loaded into sterically stabilized, solid lipid
Fanfei Meng et al.
International journal of pharmaceutics, 506(1-2), 46-56 (2016-04-21)
Many nanoparticle matrixes have been demonstrated to be efficient in brain targeting, but there are still certain limitations for them. To overcome the shortcomings of the existing nanoparticulate systems for brain-targeted delivery, a lipoprotein resembling protein-free nanostructured lipid carrier (PS80-NLC)
Clive J Petry et al.
Reproductive biology and endocrinology : RB&E, 16(1), 82-82 (2018-08-31)
Imprinted genes, which are expressed in a parent of origin-specific manner, are thought to mediate the genetic priorities of each parent in pregnancy. Recently we reported that some fetal imprinted gene variants are associated with maternal glucose concentrations and blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service