Skip to Content
Merck
All Photos(2)

Key Documents

D8269

Sigma-Aldrich

Glyceryl 1,3-distearate

≥99% (TLC)

Synonym(s):

1,3-Dioctadecanoylglycerol, 1,3-Distearin, 1,3-Distearoylglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76O5
CAS Number:
Molecular Weight:
625.02
Beilstein:
1810306
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99% (TLC)

form

powder

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3

InChI key

IZHVBANLECCAGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H O Herrmann et al.
Molecular and biochemical parasitology, 3(5), 319-325 (1981-09-01)
A medium-chain O-alkylglycerol, 1-O-[1'-14C]dodecyl-sn-glycerol, has been found to be incorporated into plasmenyl ethanolamine by Leishmania donovani promastigotes as revealed by radio gas-liquid chromatography; however, the ether bond of the administered O-alkyl-glycerol was cleaved extensively as judged from the occurrence of
R Brasseur et al.
Biochemical and biophysical research communications, 127(3), 969-976 (1985-03-29)
Phorbol esters and diacylglycerols activate protein kinase C but specific structural parameters appear to be required for the enzyme activation. We have analyzed the conformation of potent and not potent diacylglycerols and phorbol esters. The orientation of the CH20H group
Kyeong-Ok Choi et al.
Journal of food science, 85(10), 3432-3441 (2020-09-13)
Combination of oleogelators has recently started to receive scientific attention since single oleogelator may not adequately compensate for the diverse roles of solid fat in a complicated food system. In this study, grapeseed oil oleogels were prepared with candelilla wax
M C Luzardo et al.
Archives of biochemistry and biophysics, 363(1), 81-90 (1999-03-02)
The physical properties conferred to DPPC bilayers by including neoglycolipids composed by two different trisaccharides: mannose-mannose-mannose (3M) and glucose-mannose-glucose (GMG) attached to a cholesterol (cho) and a distearylglycerol (diC18) lipid moiety by a spacer were evaluated by means of the
Jarernporn Thawornprasert et al.
ACS omega, 5(32), 20021-20033 (2020-08-25)
This research focuses on the feasibility of using diesel-palm fatty acid distillate ethyl ester (PFADE)-ethanol in a direct injection diesel engine without any major modifications. Hydrous ethanol was selected for blending in diesel to produce diesohol. The palm fatty acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service