Skip to Content
Merck
All Photos(1)

Key Documents

PHR1335

Supelco

Labetalol Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Labetalol hydrochloride, 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2O3 · HCl
CAS Number:
Molecular Weight:
364.87
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP L0050000
traceable to USP 1356654

API family

labetalol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.CC(CCc1ccccc1)NCC(O)c2ccc(O)c(c2)C(N)=O

InChI

1S/C19H24N2O3.ClH/c1-13(7-8-14-5-3-2-4-6-14)21-12-18(23)15-9-10-17(22)16(11-15)19(20)24;/h2-6,9-11,13,18,21-23H,7-8,12H2,1H3,(H2,20,24);1H

InChI key

WQVZLXWQESQGIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Labetalol Hydrochloride is an adrenergic α-receptor and β-receptor blocking agent, widely used in the treatment of hypertension. It is a sensitive drug and is used quantitatively especially in myocardial infarction and unstable angina, since it decreases blood pressure more rapidly than other beta blockers.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Labetalol Hydrochloride may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1069 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of labetalol hydrochloride by kinetic spectrophotometry using potassium permanganate as oxidant
Rahman N, et al.
Journal of the Mexican Chemical Society, 55(2), 105-112 (2011)
Determination of labetalol hydrochloride in drug formulations by spectrophotometry
Rahman N, et al.
J. Chin. Chem. Soc., 54(1), 185-196 (2007)
Chitra Saran et al.
The Journal of pharmacology and experimental therapeutics (2021-11-20)
Drug-induced liver injury (DILI) is the leading cause of acute liver failure and a major concern in drug development. Altered bile acid homeostasis via inhibition of the bile salt export pump (BSEP) is one mechanism of DILI. Dasatinib, pazopanib and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service