Skip to Content
Merck
All Photos(1)

Documents

PHR1286

Supelco

Gemfibrozil

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, 2,2-Dimethyl-5-(2,5-xylyloxy)valeric acid, 5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O3
CAS Number:
Molecular Weight:
250.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 363
traceable to Ph. Eur. Y0000513
traceable to USP 1288500

API family

gemfibrozil

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1ccc(C)c(OCCCC(C)(C)C(O)=O)c1

InChI

1S/C15H22O3/c1-11-6-7-12(2)13(10-11)18-9-5-8-15(3,4)14(16)17/h6-7,10H,5,8-9H2,1-4H3,(H,16,17)

InChI key

HEMJJKBWTPKOJG-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Gemfibrozil is a cholesterol-lowering drug that effectively lowers the serum cholesterol, triglyceride, and low-density lipoprotein (LDL) levels. It also efficiently raises the serum high-density lipoprotein (HDL) levels. It is known to minimize the incidence of coronary heart diseases in humans.

Application

Gemfibrozil may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by high performance liquid chromatography (HPLC).
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0300 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of liquid chromatographic method for gemfibrozil and simvastatin in binary combination
Ashfaq M, et al.
Journal of the Chilean Chemical Society, 53(3), 1617-1619 (2008)
Gianluca Miglio et al.
British journal of pharmacology, 167(3), 641-653 (2012-05-19)
Peroxisome proliferator-activated receptor (PPAR) agonists exert anti-albuminuric effects. However, the nephroprotective effects of these drugs remain to be fully understood. We have investigated whether gemfibrozil, GW0742 and pioglitazone protect human podocytes against nutrient deprivation (ND)-induced cell death and the role
Sarah Y Skolness et al.
Environmental toxicology and chemistry, 31(11), 2615-2624 (2012-08-23)
Fibrates are a class of pharmaceuticals that indirectly modulate cholesterol biosynthesis through effects on peroxisome proliferator-activated receptors. Gemfibrozil is a fibrate that has been detected in wastewater treatment plant influents, effluents, and drinking water. The objective of the present study
C M Spencer et al.
Drugs, 51(6), 982-1018 (1996-06-01)
Gemfibrozil improves lipid and apolipoprotein profiles, particularly very low density lipoprotein (VLDL) triglyceride and high density lipoprotein (HDL) cholesterol levels, in patients with dyslipidaemia when administered at a total daily dose of 900 or 1200 mg. As demonstrated by the
Ana Maria Sierra Villar et al.
International journal of pharmaceutics, 431(1-2), 161-175 (2012-04-14)
Self-nanoemulsifying drug delivery systems of gemfibrozil were developed under Quality by Design approach for improvement of dissolution and oral absorption. Preliminary screening was performed to select proper components combination. Box-Behnken experimental design was employed as statistical tool to optimize the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service