Skip to Content
Merck
All Photos(1)

Documents

Y0000514

Gemfibrozil for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Gemfibrozil, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, 2,2-Dimethyl-5-(2,5-xylyloxy)valeric acid, 5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O3
CAS Number:
Molecular Weight:
250.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

gemfibrozil

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1ccc(C)c(OCCCC(C)(C)C(O)=O)c1

InChI

1S/C15H22O3/c1-11-6-7-12(2)13(10-11)18-9-5-8-15(3,4)14(16)17/h6-7,10H,5,8-9H2,1-4H3,(H,16,17)

InChI key

HEMJJKBWTPKOJG-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Gemfibrozil for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amir Qaseem et al.
Annals of internal medicine, 159(12), 835-847 (2013-10-23)
The American College of Physicians (ACP) developed this guideline to present the evidence and provide clinical recommendations on the screening, monitoring, and treatment of adults with stage 1 to 3 chronic kidney disease. This guideline is based on a systematic
Gianluca Miglio et al.
British journal of pharmacology, 167(3), 641-653 (2012-05-19)
Peroxisome proliferator-activated receptor (PPAR) agonists exert anti-albuminuric effects. However, the nephroprotective effects of these drugs remain to be fully understood. We have investigated whether gemfibrozil, GW0742 and pioglitazone protect human podocytes against nutrient deprivation (ND)-induced cell death and the role
Christina L Aquilante et al.
British journal of clinical pharmacology, 75(1), 217-226 (2012-05-26)
The objective of this study was to determine the extent to which the CYP2C8*3 allele influences pharmacokinetic variability in the drug-drug interaction between gemfibrozil (CYP2C8 inhibitor) and pioglitazone (CYP2C8 substrate). In this randomized, two phase crossover study, 30 healthy Caucasian
Manthena V S Varma et al.
Pharmaceutical research, 29(10), 2860-2873 (2012-05-29)
To develop physiologically based pharmacokinetic (PBPK) model to predict the pharmacokinetics and drug-drug interactions (DDI) of pravastatin, using the in vitro transport parameters. In vitro hepatic sinusoidal active uptake, passive diffusion and canalicular efflux intrinsic clearance values were determined using
A M Filppula et al.
Clinical pharmacology and therapeutics, 94(3), 383-393 (2013-05-10)
Cytochrome P450 (CYP) 3A4 is considered the most important enzyme in imatinib biotransformation. In a randomized, crossover study, 10 healthy subjects were administered gemfibrozil 600 mg or placebo twice daily for 6 days, and imatinib 200 mg on day 3, to study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service