Skip to Content
Merck
All Photos(1)

Key Documents

PHR1217

Supelco

1-Pentanol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

n-Amyl alcohol, Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730975
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1504955

vapor density

3 (vs air)

vapor pressure

1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

CofA

current certificate can be downloaded

expl. lim.

10 %, 100 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.409 (lit.)

bp

136-138 °C (lit.)

mp

−78 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCCCO

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. 1-Pentanol is a linear alcohol and a promising substitute for conventional gasoline and diesel fuels.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9030 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acid/base- and anion-controllable organogels formed from a urea-based molecular switch.
Sheng-Yao Hsueh et al.
Angewandte Chemie (International ed. in English), 49(48), 9170-9173 (2010-10-19)
Nadav Amdursky et al.
The journal of physical chemistry. A, 115(24), 6481-6487 (2011-05-19)
Time-resolved emission techniques were employed to study the nonradiative process of thioflavin-T (ThT) in 1-propanol, 1-butanol, and 1-pentanol as a function of the hydrostatic pressure. Elevated hydrostatic pressure increases the alcohol viscosity, which in turn strongly influences the nonradiative rate
Bidyut K Paul et al.
Journal of colloid and interface science, 316(2), 751-761 (2007-10-02)
The phase behaviors, interfacial composition, thermodynamic properties and structural characteristics of water-in-oil microemulsions under varied molar ratio of water to surfactant (omega) at 303 K and also by varying temperatures at a fixed omega(=40) by mixing with 1-pentanol and decane
Gizelle Cristina Bedendo et al.
Journal of chromatography. A, 1217(42), 6449-6454 (2010-09-14)
A sensitive and precise analysis using hollow fiber renewal liquid membrane (HFRLM) extraction followed by high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) is described for determination of five sulfonamides in honey samples. In this procedure, the organic solvent introduced directly
Zhisheng Wu et al.
Bioorganic & medicinal chemistry letters, 22(20), 6358-6361 (2012-09-22)
N-(rhodamine B)-deoxylactam-5-amino-1-pentanol (dRB-APOH) was designed and prepared as the chromo-fluorogenic sensor for detection of a nerve agent simulant via analyte triggered tandem phosphorylation and opening of the intramolecular deoxylactam. The successful detection of diethyl chlorophosphate suggests the utility of rhodamine-deoxylactams

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service