Skip to Content
Merck
1-Pentanol puriss. p.a., ACS reagent, ≥99.0% (GC)

1-Pentanol

1-Pentanol puriss. p.a., ACS reagent, ≥99.0% (GC)
Synonym(s):
n-Amyl alcohol, Pentyl alcohol
Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique