Skip to Content
Merck
All Photos(1)

Documents

77664

Supelco

3-Methyl-1-butanol

analytical standard

Synonym(s):

Isoamyl alcohol, Isopentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718835
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

Assay

≥98.5% (GC)

autoignition temp.

644 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.2-9 %, 100 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.406 (lit.)

bp

130 °C (lit.)

mp

−117 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-1-butanol is a potential fuel additive, which is a natural product of fungus and yeast fermentation.

Application

3-Methyl-1-butanol is used as a precursor for the synthesis of various chemicals, such as isoamyl acetate. It may be used as a reference standard for the analysis of 3-methyl-1-butanol content in honey samples using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contribution of dynamic headspace GC-MS analysis of aroma compounds to authenticity testing of honey.
Radovic SB, et al.
Food Chemistry, 72(4), 511-520 (2001)
Michael R Connor et al.
Applied and environmental microbiology, 74(18), 5769-5775 (2008-08-05)
3-Methyl-1-butanol is a potential fuel additive or substitute. Previously this compound was identified in small quantities in yeast fermentation as one of the fusel alcohols. In this work, we engineered an Escherichia coli strain to produce 3-methyl-1-butanol from glucose via
Luca Bianco et al.
PloS one, 9(10), e110377-e110377 (2014-10-11)
High-density SNP arrays for genome-wide assessment of allelic variation have made high resolution genetic characterization of crop germplasm feasible. A medium density array for apple, the IRSC 8K SNP array, has been successfully developed and used for screens of bi-parental
I H Chan et al.
Mucosal immunology, 7(4), 842-856 (2013-11-28)
Chronic inflammation has been associated with increased risk for developing gastrointestinal cancer. Interleukin-23 (IL-23) receptor signaling has been correlated with inflammatory bowel disease pathogenesis, as well as promotion of tumor growth. However, little is known about the relative potential for
Shantala Arundhati Hari Dass et al.
Molecular ecology, 23(24), 6114-6122 (2014-08-22)
Male rats (Rattus novergicus) infected with protozoan Toxoplasma gondii relinquish their innate aversion to the cat odours. This behavioural change is postulated to increase transmission of the parasite to its definitive felid hosts. Here, we show that the Toxoplasma gondii

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service