Skip to Content
Merck
All Photos(1)

Documents

PHR1161

Supelco

Procaine hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Aminobenzoic acid 2-diethylaminoethyl ester, p-Aminobenzoic acid diethylaminoethyl ester hydrochloride, Novocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2CH2CH2N(C2H5)2·HCl
CAS Number:
Molecular Weight:
272.77
Beilstein:
3917802
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1564006

API family

procaine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

155-156 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.CCN(CC)CCOC(=O)c1ccc(N)cc1

InChI

1S/C13H20N2O2.ClH/c1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h5-8H,3-4,9-10,14H2,1-2H3;1H

InChI key

HCBIBCJNVBAKAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Procaine hydrochloride is a synthetic compound that belongs to the class of local anesthetic drugs. It has a structure similar to the natural compounds that participate in nerve impulse transmission. It shows interaction with the cellular membranes causing membrane lipids to expand, membrane-bound calcium to displace and change the structure of the proteins that are responsible for regulating the cell permeability and shape.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Procaine hydrochloride can be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by electrochemical, chemiluminescence, and spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7177 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of procaine, benzocaine and tetracaine by sequential injection analysis with permanganate-induced chemiluminescence detection
Pasekova H and Polasek M
Talanta, 52(1), 67-75 (2000)
Improved therapeutic index of cisplatin by procaine hydrochloride
Esposito M, et al.
Journal of the National Cancer Institute, 82(8), 677-684 (1990)
Colorimetric Determination of Procaine hydrochloride in Pharmaceutical Preparations using Diazotization Coupling Reactions
Al-Uzri WA and Al-Heeti MI
International Journal of PharmTech Research, 8(1), 1042-1042 (2015)
Effect of the presence of ?-cyclodextrin on the solution behavior of procaine hydrochloride. Spectroscopic and thermodynamic studies
Merino C, et al.
Langmuir, 16(4), 1557-1565 (2000)
Batch and flow-injection spectrophotometric determination of procaine HCl in pharmaceutical preparations via using diazotization and coupling reaction
Al-Ward HS, et al.
Baghdad Science Journal, 9(3), 521-531 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service