Skip to Content
Merck
All Photos(1)

Key Documents

PHR1249

Supelco

Phenacetin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-Acetyl-p-phenetidin, 4′-Ethoxyacetanilide, N-(4-Ethoxyphenyl)acetamide, p-Acetophenetidide, Acetophenetidin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OC2H5
CAS Number:
Molecular Weight:
179.22
Beilstein:
1869238
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1513005

API family

phenacetin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

133-136 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCOc1ccc(NC(C)=O)cc1

InChI

1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)

InChI key

CPJSUEIXXCENMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Phenacetin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Substrate of CYP1A2 and CYP2D6.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9053 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparative bioavailability study of different aspirin formulations using on-line multidimensional chromatography.
Sagar KA and Smyth MR
Journal of Pharmaceutical and Biomedical Analysis, 21(2), 383-392 (1999)
Some applications of bonded-phase high-performance liquid chromatography to the analysis of pharmaceutical formulations.
Cox GB, et al.
Analytica Chimica Acta, 92(2), 345-352 (1977)
Validation of a HPLC quantification of acetaminophen, phenylephrine and chlorpheniramine in pharmaceutical formulations: capsules and sachets.
Marin A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(4), 701-714 (2002)
Analgesic nephropathy: a reassessment of the role of phenacetin and other analgesics.
L F Prescott
Drugs, 23(1-2), 75-149 (1982-01-01)
S P Clissold
Drugs, 32 Suppl 4, 46-59 (1986-01-01)
Since their synthesis in the late 1800s paracetamol (acetaminophen) and phenacetin have followed divergent pathways with regard to their popularity as mild analgesic/antipyretic drugs. Initially, paracetamol was discarded in favour of phenacetin because the latter drug was supposedly less toxic.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service