Skip to Content
Merck
All Photos(2)

Key Documents

O1383

Sigma-Aldrich

Oleic acid

BioReagent, suitable for cell culture

Synonym(s):

cis-9-Octadecenoic acid, Elainic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
282.46
Beilstein:
1726542
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.75

biological source

palm oil

Quality Level

vapor pressure

1 mmHg ( 176 °C)

product line

BioReagent

Assay

≥99% (GC)

form

viscous liquid

technique(s)

cell culture | mammalian: suitable

refractive index

n20/D 1.459 (lit.)

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

solubility

ethanol: soluble

density

0.89 g/mL at 25 °C (lit.)
0.89 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

ZQPPMHVWECSIRJ-KTKRTIGZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Oleic acid is a mono-unsaturated omega-9 fatty acid created by dehydrogenation of stearic acid. It is then further dehydrogenated and elongated to create members of the n-9 family of fatty acids. Oleic acid is not an essential fatty acid (EFA); however, providing it to cells in culture reduces energy demand.
Oleic acid mediates complete oxidation of fatty acid in sirtuin1- proliferator-activated receptor γ coactivator−1α (SIRT1-PGC-1α) signalling pathway. Oleic acid shows anti-inflammatory properties related to atherosclerosis and insulin resistant state. Oleic acid inhibits permeability in gap junctions and promotes glucose uptake in astrocytes. Oleic acid aids in neural cell proliferation by increasing glucose uptake and by protein kinase C (PKC) activation. Oleic acid is essential for axonal growth and neuronal clustering.

Application

Oleic acid has been used:
  • in fatty acid oxidation rate experiments in myotubes and in Sirt1 enzyme assay
  • to determine quantitatively its effect on lipid droplets by optical diffraction tomography
  • as a fatty acid source in the medium for primary hepatocyte culture

Packaging

Sealed ampule.

Caution

Unsaturated fatty acids are subject to oxidation and peroxidation. Consider complexed fatty acid supplements.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Three-dimensional label-free imaging and quantification of lipid droplets in live hepatocytes
Kim K, et al.
Scientific Reports, 6(10), 36815-36815 (2016)
Beneficial effect of fish oil on human brain (2009)
The role of proliferator-activated receptor gamma coactivator-1alpha in the fatty-acid-dependent transcriptional control of interleukin-10 in hepatic cells of rodents
Morari J, et al.
Metabolism, Clinical and Experimental, 59(2), 215-223 (2010)
Oleic acid stimulates complete oxidation of fatty acids through protein kinase A-dependent activation of SIRT1-PGC1alpha complex
Lim JH, et al.
The Journal of Biological Chemistry, 288(10), 7117-7126 (2013)
Marta Estrader et al.
Nanoscale, 7(7), 3002-3015 (2015-01-21)
The intimate relationship between stoichiometry and physicochemical properties in transition-metal oxides makes them appealing as tunable materials. These features become exacerbated when dealing with nanostructures. However, due to the complexity of nanoscale materials, establishing a distinct relationship between structure-morphology and

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service