Skip to Content
Merck
All Photos(1)

Key Documents

NIST875

Cupro-nickel

NIST SRM 875, 10% (CDA 706) “DOPED”

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 100 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This material is in the form of small granules prepared by water atomization and is intneded for use in chemical and instrumental methods of analysis. For more information, please refer to the SDS and COA.

SRM 875_cert

SRM 875_SDS

Other Notes

Certified for the analytes listed below.
Copper (Cu), Nickel (Ni), Iron (Fe), Zinc (Zn), Lead (Pb), Manganese (Mn), Antimony (Sb), Tin (Sn), Phosphorus (P), Bismuth (Bi), Cadmium (Cd), Selenium (Se)

Reference/Informational value is provided for the following analytes.
Oxygen (O), Hydrogen (H), Carbon (C), Sulfur (S), Magnesium (Mg), Arsenic (As), Silicon (Si), Titanium (Ti), Tellurium (Te)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service