Skip to Content
Merck
All Photos(1)

Key Documents

N0400000

Neomycin sulphate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H46N6O13 · xH2SO4
CAS Number:
Molecular Weight:
614.64 (free base basis)
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

neomycin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[S](=O)(=O)(O)O.N[C@H]1[C@H](O[C@@H]([C@H]([C@@H]1O)O)CN)O[C@H]2[C@@H]([C@H]([C@@H](C[C@@H]2N)N)O)O[C@@H]3O[C@@H]([C@H]([C@H]3O)OC4O[C@H]([C@H]([C@@H]([C@H]4N)O)O)CN)CO

InChI

1S/C23H46N6O13.H2O4S/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;1-5(2,3)4/h5-23,30-36H,1-4,24-29H2;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22?,23+;/m1./s1

InChI key

OIXVKQDWLFHVGR-GQTDVWSESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Neomycin sulphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Almut G Winterstein et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 148(2), 277-283 (2012-12-25)
Use of neomycin eardrops in nonintact tympanic membranes (NITMs) due to tympanic membrane (TM) perforation or tympanostomy tubes (TTs) is controversial because of the potential for ototoxicity. We sought to compare the risk of sensorineural hearing loss (SNHL) in patients
Venkatareddy Udumula et al.
Bioorganic & medicinal chemistry letters, 23(6), 1671-1675 (2013-02-19)
Aminoglycoside represents a class of versatile and broad spectrum antibacterial agents. In an effort to revive the antibacterial activity against aminoglycoside resistant bacteria, our laboratory has developed two new classes of aminoglycoside, pyranmycin and amphiphilic neomycin (NEOF004). The former resembles
K Stypulkowska et al.
Journal of pharmaceutical and biomedical analysis, 76, 207-214 (2013-01-23)
A new, simple and repeatable liquid chromatographic method with charged aerosol detection (LC-CAD) for determination of neomycin and related substances has been developed. Analysis of neomycin or other aminoglycosides is problematic due to a lack of chromophores. Universal response of
Jakob Torp Madsen et al.
Contact dermatitis, 68(2), 94-97 (2013-01-08)
Background. Two readings of patch test reactions are recommended. Objectives. To evaluate the outcome of a second patch test reading of TRUE Test® allergens on D6/7 in relation to negative or doubtful reactions on D3/4. Methods. This was a retrospective
Vijayalakshmi Ghosh et al.
Colloids and surfaces. B, Biointerfaces, 105, 152-157 (2013-01-30)
Microemulsions are thermodynamically stable isotropic systems comprising of oil, surfactant and water. Cinnamon oil (Cinnamomum zeylanicum) microemulsion was formulated using non-ionic surfactant Tween 20 and water. With oil to surfactant (v/v) ratio of 1:4, cinnamon oil microemulsion (CMF4) was formulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service