Skip to Content
Merck
All Photos(1)

Key Documents

P2400000

Polymyxin B sulfate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

polymyxin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[S](=O)(=O)(O)O.N1CCC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C1=O)CCO)CCN)CCN)CC(C)C)Cc2ccccc2)CCN)NC(=O)C(NC(=O)C(NC(=O)C(NC=O)CCN)CCO)CCN

InChI

1S/C48H82N16O13.H2O4S/c1-27(2)24-37-47(76)59-32(11-19-52)41(70)56-31(10-18-51)43(72)61-35(14-22-65)39(68)54-21-13-34(45(74)57-33(12-20-53)44(73)64-38(48(77)63-37)25-28-6-4-3-5-7-28)60-42(71)30(9-17-50)58-46(75)36(15-23-66)62-40(69)29(8-16-49)55-26-67;1-5(2,3)4/h3-7,26-27,29-38,65-66H,8-25,49-53H2,1-2H3,(H,54,68)(H,55,67)(H,56,70)(H,57,74)(H,58,75)(H,59,76)(H,60,71)(H,61,72)(H,62,69)(H,63,77)(H,64,73);(H2,1,2,3,4)

InChI key

HNDFYNOVSOOGDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Polymyxin B sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Nakamura et al.
The American journal of emergency medicine, 31(5), 893-893 (2013-02-13)
Septic patients often have low cardiac output. Some of them present severe cardiac dysfunction such as septic cardiomyopathy. However, no well-known and effective treatment for septic cardiomyopathy exists. The effect of endotoxin adsorption by polymyxin B–immobilized fiber column–direct hemoperfusion (PMX-DHP)
Lee Sun New et al.
Analytical and bioanalytical chemistry, 405(4), 1407-1415 (2012-11-28)
With increasing multidrug resistance coupled to a poor development pipeline, clinicians are exploring antimicrobial combinations to improve treatment outcomes. In vitro hollow-fiber infection model (HFIM) is employed to simulate human in vivo drug clearance and investigate pharmacodynamic synergism of antibiotics.
A Bonura et al.
Allergy, 68(3), 297-303 (2013-01-22)
The major allergens in Parietaria pollen, Par j 1 and Par j 2, have been identified as lipid transfer proteins. The family of the Par j 1 allergens is composed of two isoforms, which differ by the presence of a
Naoki Sawa et al.
Clinical nephrology, 79(6), 463-470 (2012-12-21)
Vasopressin and direct hemoperfusion with an immobilized polymyxin B column (PMX) have emerged recently as treatments for septic shock. This study assessed the impact of these two treatments on the survival of patients with septic shock. A retrospective, matched cohort
Marco M Domingues et al.
Biopolymers, 98(4), 338-344 (2012-11-30)
Antibiotic resistance is an increasingly severe health problem. Antimicrobial peptides (AMPs) are being developed in order to overcome this problem, due lower bacterial resistance. Polymyxin B is an AMP with bactericidal effect on Gram-negative bacteria due to its high affinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service