Skip to Content
Merck
All Photos(1)

Key Documents

N0392

Supelco

Nordoxepin hydrochloride

analytical standard, ≥98.0% (TLC), powder

Synonym(s):

11(6H)-(3-[Methylamino]propylidene)dibenz[b,e]oxepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO · HCl
CAS Number:
Molecular Weight:
301.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (TLC)

form

powder

technique(s)

HPLC: suitable

color

white to yellow

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CNCC\C=C1/c2ccccc2COc3ccccc13

InChI

1S/C18H19NO.ClH/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18;/h2-5,7-11,19H,6,12-13H2,1H3;1H/b16-10+;

InChI key

GNPPEZGJRSOKRE-QFHYWFJHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Doxepin metabolite.

Features and Benefits

Shelf-life of the powder is at least 10 years.

Other Notes

Mixture of cis- and trans-isomers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C W Harrell et al.
Electrophoresis, 19(5), 712-718 (1998-06-18)
The separation of seven structurally similar antidepressant drugs (amitriptyline, nortriptyline, imipramine, desipramine, protriptyline, doxepin, and nordoxepin) was achieved in under 15 min using a novel nonionic micelle polymer, poly(n-undecyl-alpha-D-glucopyranoside) (PUG) by use of capillary zone electrophoresis (CZE). Systematic studies with
J Yan et al.
Journal of chromatography. B, Biomedical sciences and applications, 691(1), 131-138 (1997-03-28)
Doxepin is a tricyclic antidepressant marketed as an irrational mixture of cis- and trans-geometric isomers in the ratio of 15:85. A convenient high-performance liquid chromatographic (HPLC) procedure for simultaneous quantitation of geometric isomers of doxepin and N-desmethyldoxepin in plasma and
Antonia Gronewold et al.
Forensic science international, 190(1-3), 74-79 (2009-06-16)
Body fluids and tissues in eight doxepin (Dox)-related deaths were investigated in order to prove whether the individual concentration of Dox, the concentration sum of parent drug and its active metabolite N-desmethyldoxepin (NDox) or the concentration ratio Dox/Ndox valuably contribute
Robert Schomburg et al.
Journal of neural transmission (Vienna, Austria : 1996), 118(4), 641-645 (2011-02-26)
Doxepin--like other antidepressant drugs (ADs)--shows a variable antidepressant effect in clinical practice. The cause for this variability is as yet unclear; however, pharmacokinetic factors such as the variable permeability of doxepin into the cerebrospinal fluid (CSF), may contribute to the
K K Midha et al.
European journal of clinical pharmacology, 42(5), 539-544 (1992-01-01)
Commercial preparations of the tricyclic anti-depressant doxepin contain 15% of the more active cis-doxepin and 85% of the trans-isomer. The single dose pharmacokinetics of doxepin and its major metabolite N-desmethyldoxepin were examined in 30 healthy young men. Results for total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service