Skip to Content
Merck

N7261

Nortriptyline hydrochloride

≥98% (HPLC), norepinephrine and serotonin inhibitor, powder

Synonym(s):

3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-1-propanamine hydrochloride, Desmethylamitriptyline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H21N · HCl
CAS Number:
Molecular Weight:
299.84
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
212-973-0
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nortriptyline hydrochloride, ≥98% (HPLC), powder

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

ethanol: 100 mg/mL, H2O: soluble

originator

Eli Lilly

SMILES string

Cl.CNCC\C=C1\c2ccccc2CCc3ccccc13

InChI

1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20H,6,12-14H2,1H3;1H

InChI key

SHAYBENGXDALFF-UHFFFAOYSA-N

Biochem/physiol Actions

Tricyclic antidepressant that is a more potent inhibitor of the norepinephrine transporter (Ki = 3.4 nM) than the serotonin transporter (Ki = 161 nM). 5-HT2 serotonin receptor antagonist.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Nortriptyline hydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library